Skip to Content
Merck
All Photos(2)

Key Documents

ABT333

Sigma-Aldrich

Anti-IL-1RAcP Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

IL-1RAcP, C3orf13, IL1R3, Interleukin-1 receptor accessory protein, IL-1 receptor accessory protein, IL-1RAcP, Interleukin-1 receptor 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€459.00

€459.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€459.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€459.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IL1RAP(3556)

General description

Interleukin-1 receptor accessory protein (IL-1RAcP), also known as Interleukin-1 receptor 3 (IL-1R-3, IL-1R3) forms a complex with IL-1RI in response to IL-1 treatment. The IL-1 receptor-associated kinase (IRAK), which mediates activation of NF-kB inducing kinae (NIK) and of NF-kB, recruites to the IL-1R complex through IL-1 RAcP. IL-1 activation of stress-activated protein kinase and of acid sphingomyelinase also requires IL-1RAcP. Like IL-1RI, IL-1RAcP subunit is essential for IL-1 mediated cellular response. IL-1RAcP is expressed in many tissues.

Specificity

This antibody recognizes the C-terminus of IL-1RAcP.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human IL-1RAcP.

Application

Detect Interleukin Receptor using this rabbit polyclonal antibody, Anti-IL-1RAcP Antibody validated for use in western blotting & ICC.
Immunocytochemistry Analysis: 5 ug/mL from a representative lot detected IL-1RAcP in HeLa cells.
Research Category
Cell Structure
Research Sub Category
Cytokines & Cytokine Receptors

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected IL-1RAcP in 15 µg of HeLa cell lysate.

Target description

~66 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haram Park et al.
The EMBO journal, 39(11), e104150-e104150 (2020-04-30)
Alternative splicing regulates trans-synaptic adhesions and synapse development, but supporting in vivo evidence is limited. PTPδ, a receptor tyrosine phosphatase adhering to multiple synaptic adhesion molecules, is associated with various neuropsychiatric disorders; however, its in vivo functions remain unclear. Here, we show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service