Skip to Content
Merck
All Photos(2)

Key Documents

ABT318

Sigma-Aldrich

Anti-Protocadherin 19 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protocadherin-19

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€408.00

€408.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PCDH19(57526)

General description

Protocadherin-19, also known as Potential calcium-dependent cell-adhesion protein (PCDH19), is a single-pass type I membrane protein that is moderately expressed in all regions of the brain and ovary, and has low expression in other tissues. Defects in PCDH19 cause epileptic encephalopathy, early infantile type 9 (EIEE9), also known as epilepsy female-restricted with mental retardation.

Immunogen

Epitope: Cytoplasmic domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human Protocadherin 19.

Application

Detect Protocadherin 19 using this Anti-Protocadherin 19 Antibody validated for use in IHC (Paraffin).
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected Protocadherin 19 in mouse hypothalamus and hind brain tissues, whereas a 1:2,000 dilution from a representative lot detected Protocadherin 19 in human cerebellum tissue.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Immunohistochemistry in human cerebral cortex tissue.

Immunohistochemistry Analysis: A 1:500 dilution of this antibody detected Protocadherin 19 in human cerebral cortex tissue.

Target description

126 kDa calculated

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohui Lv et al.
Nature communications, 10(1), 3946-3946 (2019-09-04)
Cerebral cortex expansion is a hallmark of mammalian brain evolution; yet, how increased neurogenesis is coordinated with structural and functional development remains largely unclear. The T-box protein TBR2/EOMES is preferentially enriched in intermediate progenitors and supports cortical neurogenesis expansion. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service