Skip to Content
Merck
All Photos(2)

Key Documents

ABT282

Sigma-Aldrich

Anti-phospho-NUMA1 Antibody (Thr2055)

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear mitotic apparatus protein 1, NuMA protein, SP-H antigen

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€446.00

€446.00


Estimated to ship on05 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on05 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), bovine (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr2055)

Gene Information

human ... NUMA1(4926)

General description

NUMA1 or NUMA protein is an interesting and major component of the nuclear matrix of eukaryotic cells. NUMA1 is a highly abundant protein component that appears to serve a non-structural role but is needed for mitotic spindle poles and centrosome formation as well as a linker of microtubules. These functions are important for proper cytokinesis and the setup of the proper polarity axis during cell division. NUMA1 resides in the nuclear matrix during interphase. NUMA1 dissociates from condensing chromosomes during early prophase, and relocates to the spindle poles via a dynein/dynamin complex, and it remains there until the anaphase onset. Thus NUMA1 then can be found in the nucleus during cell rest and cytoplasmically and centrosome localized during cytokinesis. NUMA1 phosphorylation occurs by CDK1 and phosphorylation increases solubility and promotes association with dynein and subsequent translocation to the spindle poles.

Immunogen

Epitope: Tail Region
KLH-conjugated linear peptide corresponding to the tail region of human NUMA1 phosphorylated at Thr2055.

Application

Peptide Inhibition Analysis: A 1:500 dilution from a representative lot blocked NUMA1 phosphorylated at Thr2055 in Hydroxyurea treated HeLa nuclear extract.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-phospho-NUMA1 Antibody (Thr2055) is validated for use in Western Blotting and Peptide Inhibition Assay for the detection of phospho-NUMA1 (Thr2055).

Quality

Evaluated by Western Blotting in HeLa nuclear extract and Hydroxyurea treated HeLa nuclear extract.

Western Blotting Analysis: A 1:500 dilution of this antibody detected NUMA1 phosphorylated at Thr2055 in 10 µg of HeLa nuclear extract and Hydroxyurea treated HeLa nuclear extract.

Target description

~230 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service