Skip to Content
Merck
All Photos(2)

Key Documents

ABT277

Sigma-Aldrich

Anti-Galectin-7 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Galectin-7, Gal-7, HKL-14, PI7, p53-induced gene 1 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€306.00

€306.00


Estimated to ship on25 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€306.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€306.00


Estimated to ship on25 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), primate (based on 100% sequence homology), monkey (based on 100% sequence homology), baboon (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Galectins are a family of carbohydrate binding proteins. Galectin-7 was initially isolated from human keratinocytes as a novel member of S-lectin family that was responsive to retinoic acid but not detectable in epidermal tumors or squamous carcinomas. It is an early transcriptional target of the tumor suppressor P53 and inducible by UV radiation as well. Galectin-7 is also an enhancer of apoptosis, whereas galectin-1 induces tumorigenicity and resistance to apoptosis in several types of cancers. Recent research demonstrates that Galectin-7 acts as an adhesion molecule during implantation, and increased expression of Galectin-7 is associated with miscarriage, thus Galectin-7 is also being studied as a possible biomarker for miscarriage prediction.

Immunogen

KLH-conjugated beta-galactose binding region corresponding to human Galectin-7.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Galectin-7 in human skin and human larynx tissue.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-Galectin-7 Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of Galectin-7.

Quality

Evaluated by Western Blotting in human skin tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Galectin-7 in 10 µg of human skin tissue lysate.

Target description

~16 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service