Skip to Content
Merck
All Photos(2)

Key Documents

ABT1388

Sigma-Aldrich

Anti-Phospho-Lamin A/C (Ser390)

from rabbit

Synonym(s):

Prelamin A/C, Renal carcinoma antigen NY-REN-32

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€171.00
200 μL
€403.00

€171.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 μL
€171.00
200 μL
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€171.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

phosphorylation (pSer390)

Gene Information

human ... LMNA(4000)

General description

Prelamin-A/C (UniProt: P02545) is encoded by the LMNA (also known as LMN1) gene (Gene ID: 4000) in human. Prelamin-A/C is subsequently cleaved into Lamin A/C. Lamins are components of the nuclear lamina that provides a framework for the nuclear envelope and interact with chromatin. Prelamin-A/C

Is cleaved to generate Lamin A/C. Farnesylation of prelamin-A/C facilitates nuclear envelope targeting and subsequent cleavage by ZMPSTE24/FACE1 to remove the farnesyl group produces mature Lamin-A/C that is inserted into the nuclear lamina. Lamin A and C are present in equal amounts in the lamina of mammals and they play an important role in nuclear assembly, chromatin organization, nuclear membrane and telomere dynamics. Lamins are shown to be essential for normal development of peripheral nervous system and skeletal muscle and for muscle satellite cell proliferation. Lamins also prevent fat infiltration of muscle and bone marrow, helping to maintain the volume and strength of skeletal muscle and bone. Phosphorylation of Lamins is reported to occur continuously throughout all interphase periods and takes place mainly on the assembled lamina. Phosphorylation of the major polypeptides of the lamina induces laminar disassembly during mitosis. Phosphorylated Lamin-A/C localizes to nucleoplasm. Lamin A/C undergoes phosphorylation at multiple sites and one of the best characterized phosphorylation sites is on Serine 22 and it is phosphorylated during interphase. Phosphorylation of Serine 22 stabilizes Lamin A/C. Overexpression of Lamin-A is shown to result in greater phosphorylation of Serine 22 and 390 and Lamin A/C knockdowns display reduced phosphorylation at both sites, which helps in maintaining the integrity of the diminished lamina. Mutations in LMNA gene can cause Emery-Dreifuss muscular dystrophy 2 and 3, which are characterized by weakness and atrophy of muscle without involvement of the nervous system and cardiac conduction defects. Some mutations have also been linked to familial Lipodystrophy that leads to the loss of subcutaneous adipose tissue in the lower parts of the body and accumulation of adipose tissue in the face and neck. (Ref.: Buxboim, A., et al. (2014). Curr. Biol. 24(16): 1909-1917).

Specificity

This rabbit polyclonal antibody detects human Lamin A/C phosphorylated on serine 390.

Immunogen

KLH-conjugated linear peptide corresponding to 11 amino acids from human Lamin A/C surrounding phosphorylated Serine 390.

Application

Anti-Phospho-Lamin A/C (Ser390), Cat. No. ABT1388, is a rabbit polyclonal antibody that detects Lamin A/C phosphorylated on Serine 390 and has been tested for use in Western Blotting and Peptide Inhibition Assay.
Peptide Inhibition Analysis: A 1:500 dilution from a representative lot was used with A549 cells (specific for Lamin A _C phosphorylation) for peptide block analysis.

Quality

Evaluated by Western Blotting in A549 cells.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Phospho-Lamin A/C (Ser390) in A549 cells (specific for Lamin A/C phosphorylation).

Target description

~75 kDa and 65 kDa observed; 74.14 and 65.14 kDa calculated for Lamin A and C, respectively. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-Rong Fan et al.
iScience, 26(6), 106992-106992 (2023-06-28)
Nuclear deformation has been observed in some cancer cells for decades, but its underlying mechanism and biological significance remain elusive. To address these questions, we employed human lung cancer A549 cell line as a model in context with transforming growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service