Skip to Content
Merck
All Photos(1)

Key Documents

ABT11

Sigma-Aldrich

Anti-Fibroblast activation protein alpha (FAP) Antibody

from rabbit

Synonym(s):

fibroblast activation protein, alpha, fibroblast activation protein, alpha subunit, integral membrane serine protease, seprase, 170 kDa melanoma membrane-bound gelatinase, FAP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€446.00

€446.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€446.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€446.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FAP(2191)

General description

Fibroblast activation protein alpha (FAP), also known as Seprase, is a 97 kDa, type II cell surface glycoprotein belonging to the serine protease family. Although FAP is considered fibroblast specific, it is not found in normal healthy fibroblasts. FAP is most commonly found in stromal, tumor fibroblasts, and is likely involved in ECM pericellular proteolysis, endothelial cell migration and invasion into the ECM. The presence of FAP in human epithelial carcinoma is increased, particularly in breast, colorectal, lung, and ovarian carcinomas. FAP is also transiently observed in developing cells, or in tissues that are undergoing wound healing or suffering from chronic inflammatory conditions. Due to its specific distribution both to tumor fibroblasts and to inflammed tissue, FAP makes a promising target for research in tumor-stromal studies and in therapeutic studies.

Specificity

The antibody recognizes the extracellular domain of Fibroblast activation protein alpha

Immunogen

Epitope: Extracellular domain
Histidine-tagged recombinant protein corresponding to human FAP.

Application

Detect Fibroblast activation protein alpha (FAP) using this Anti-Fibroblast activation protein alpha (FAP) Antibody validated for use in WB, IH.
Immunohistochemistry: A 1:750 dilution of a previous lot of FAP antibody was used to immunohistochemically stain frozen tissue sections of HEK-FAP or HT-29 xenograft tumors from C.B17 mice (Cheng, 2002).

Inhibition: Inhibition of dipeptidyl peptidase activity with a previous lot of FAP antibody on HT-29 xenograft tumors was demonstrated by an independent laboratory (Cheng, 2002).
Research Category
Cell Structure

Apoptosis & Cancer
Research Sub Category
ECM Proteins

Tumor Markers

Quality

Evaluated by Western Blot in HEK-FAP xenograft tumor lysate.

Western Blot Analysis: 2.5 µg/mL of this antibody detected FAP in 10 µg of HEK-FAP xenograft tumor lysate.

Target description

~ 90 kDa observed MW. Two isoforms of the Fibroblast activation protein exist at calculated MWs of 88kDa and 27 kDa It is possible that both bands may be obserbved in certain cell lines.

Physical form

Ammonium Sulfate
Format: Purified
Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK-FAP xenograft tumor lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marian C Okondo et al.
Nature chemical biology, 13(1), 46-53 (2016-11-08)
Val-boroPro (Talabostat, PT-100), a nonselective inhibitor of post-proline cleaving serine proteases, stimulates mammalian immune systems through an unknown mechanism of action. Despite this lack of mechanistic understanding, Val-boroPro has attracted substantial interest as a potential anticancer agent, reaching phase 3
Marcus Ruscetti et al.
Cell, 181(2), 424-441 (2020-04-03)
KRAS mutant pancreatic ductal adenocarcinoma (PDAC) is characterized by a desmoplastic response that promotes hypovascularity, immunosuppression, and resistance to chemo- and immunotherapies. We show that a combination of MEK and CDK4/6 inhibitors that target KRAS-directed oncogenic signaling can suppress PDAC
Samantha Vh Bayer et al.
eLife, 8 (2019-05-31)
Biomechanical changes in the tumor microenvironment influence tumor progression and metastases. Collagen content and fiber organization within the tumor stroma are major contributors to biomechanical changes (e., tumor stiffness) and correlated with tumor aggressiveness and outcome. What signals and in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service