Skip to Content
Merck
All Photos(2)

Key Documents

ABS989

Sigma-Aldrich

Anti-E6AP Antibody

serum, from rabbit

Synonym(s):

Ubiquitin-protein ligase E3A, E6AP ubiquitin-protein ligase, Human papillomavirus E6-associated protein, Oncogenic protein-associated protein E6-AP, Renal carcinoma antigen NY-REN-54

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€408.00

€408.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UBE3A(7337)

General description

E6AP/UB3A is an interesting E3 ubiquitin protein ligase with newly discovered alternative functions as well. It is the founding member of the HECT (Homologous to E6AP C terminus) family of ubiquitin ligases and recent research is shedding new light on its influence in health and disease. Ubiquitin substrates for E6AP/UB3A ligase include RAD23A/B, MCM7 (items involved in DNA replication), Annexin A1, PML tumor suppressor and the cell cycle regulator CDKN1B. E6AP also catalyzes the high risk human papilloma virus E6 mediated ubiquitination of p53, which contributes to the neoplastic progression of cells infected by these viruses. E6AP has also been identified as a steroid hormone receptor transcriptional coactivator, and E6AP has a role in numerous diseases including cancers and neurological syndromes. Additionally, E6AP/UB3A is developmentally important for adipogenesis, granulocyte maturation and synaptic processes, where it acts as a down-regulator. IE6AP/UBE3A protein is active as a trimer of E6AP subunits and widely expressed.

Immunogen

a small portion of the full length protein expressed as a recombinant

Application

Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-E6AP Antibody is validated for use in Western Blotting and Immunoprecipitation for the detection of E6AP.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected E6AP in 10 µg of A431, C2C12, L6, mouse brain, and PC3 cell lysate.
Western Blotting Analysis: A representative lot detected E6AP in HeLa cell lysate (Talis, A. L., et al. (1998). JBC. 273:6439-6445).
Immunoprecipitation Analysis: A representative lot immunoprecipitated E6AP in HeLa cell lysate (Wynn, H. K., et al. (2000). Journal of Virology. 74(14):6408-6417).

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected E6AP in 10 µg of A431 cell lysate.

Target description

~100 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service