Skip to Content
Merck
All Photos(1)

Key Documents

ABS72

Sigma-Aldrich

Anti-WAS protein family homolog 1 (CT) Antibody

from rabbit

Synonym(s):

WAS protein family homolog 1, family with sequence similarity 39, member E, CXYorf1-like protein on chromosome 2, Protein FAM39E

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€381.00

€381.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€381.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€381.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Wiskott-Aldrich Syndrome Protein and SCAR Homolog (WASH) is a novel protein belonging to the WASP family, a group of nucleation promoting factors (NFPs) that activate a critical component in endocytosis, Arp 2/3. WASH contains three specific domains: a C-terminal VCA domain, a proline-rich domain, and an N terminal region containing two WASH homolog regions, WHD1 and WHD2. Although WASH has yet to be completely characterized, it is known to function as an NPF at the endosomal surface, where it is involved in the recruitment and subsequent activation of Arp2/3 in order to initiate actin polymerization. Therefore, WASH has a crucial role in endosomal sorting as a mediator of tubule fission resulting from Arp2/3 activation.

Specificity

This antibody recognizes WAS protein family homolog 1 at the C-terminus.

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to human WAS protein family homolog 1 at the C-terminus.

Application

Anti-WAS protein family homolog 1 (C-terminus) Antibody detects level of WAS protein family homolog 1 (C-terminus) & has been published & validated for use in WB.
Research Category
Signaling

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected WAS protein family homolog 1 on 10 µg of HeLa cell lysate.

Target description

~ 60 kDa observed. The calculated molecular weight is 50 kDa

Physical form

Format: Purified
Protein G
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María Tello-Lafoz et al.
Traffic (Copenhagen, Denmark), 18(8), 491-504 (2017-05-10)
T Lymphocyte recognition of antigens leads to the formation of a highly organized structure termed immune synapse (IS) by analogy with the neuronals synapse. Sorting nexin 27 (SNX27) controls the endosomal traffic of PSD95, Dlg1, ZO-1 (PDZ) domain-interacting proteins, and
Benjamin J Tyrrell et al.
Pigment cell & melanoma research, 29(5), 559-571 (2016-07-09)
The five-subunit WASH complex generates actin networks that participate in endocytic trafficking, migration and invasion in various cell types. Loss of one of the two subunits WASH or strumpellin in mice is lethal, but little is known about their role
Eszter Zavodszky et al.
Nature communications, 5, 3828-3828 (2014-05-14)
Endosomal protein sorting controls the localization of many physiologically important proteins and is linked to several neurodegenerative diseases. VPS35 is a component of the retromer complex, which mediates endosome-to-Golgi retrieval of membrane proteins such as the cation-independent mannose 6-phosphate receptor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service