Skip to Content
Merck
All Photos(1)

Documents

ABS526

Sigma-Aldrich

Anti-Perilipin-4, N-term Antibody

0.1 mg/mL, from rabbit

Synonym(s):

Perilipin-4, Adipocyte protein S3-12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

concentration

0.1 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Plin4(57435)

General description

Perilipin-4, also known as PLIN4, or Adipocyte protein S3-12, and encoded by the gene PLIN4/KIAA1881, plays a role in triacylglycerol metabolism in adipocytes and governs the biogenesis of lipid droplets. Perilipin-4 is also necessary for proper adipocyte maturation. Because of its role in lipid droplet formation and their involvement in the formation of macrophagic foam cells, Perilipin 4 and its other family members are seen as key players in atherogenesis : The development of atherosclerotic plaques in the blood vessels. Thus Perilipin-4 and its family members are a focus of much research from metabolic syndrome to vascular disease. Perilipin-4 is a membrane protein found in adipocyte precursors as well as mature fat cells. Interestingly too, certain polymorphims in PLIN4 are associated with height in both males and females and suggests that PLIN4 and its other family members may be involved in general growth and development as well as adipogenesis.

Immunogen

Epitope: N-terminus
Linear peptide corresponding to the N-terminus of mouse Perilipin-4.

Application

Immunohistochemistry Analysis: A representative lot detected Perilipin-4 in mouse fat tissue (Wolins, N. E., et al. (2003). JBC. 278(39):37713-37721).
Immunocytochemistry Analysis: A representative lot detected Perilipin-4 in 3T3_L1 cells (Wolins, N. E., et al. (2003). JBC. 278(39):37713-37721).
Western Blotting Analysis: A representative lot detected Perilipin-4 in White Adipose Tissue (WAT) (Wolins, N. E., et al. (2003). JBC. 278(39):37713-37721).
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-Perilipin-4 antibody, N-term is validated for use in western blotting, IHC & ICC for the detection of Perilipin-4.

Quality

Evaluated by Western Blotting in NIH/3T3-L1 cell lysate.

Western Blotting Analysis: A 1:1,000 of this antibody detected Perilipin-4 in 10 µg of NIH/3T3-L1 cell lysate.

Target description

~ 160 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS without preservatives with 10% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karuna Irungbam et al.
Laboratory investigation; a journal of technical methods and pathology, 100(3), 454-465 (2019-10-02)
The endocannabinoid (EC) system has been implicated in the pathogenesis of several metabolic diseases, including nonalcoholic fatty liver disease (NAFLD). With the current study we aimed to verify the modulatory effect of endocannabinoid receptor 1 (CB1)-signaling on perilipin 2 (PLIN2)-mediated
Anastasia Asimakopoulou et al.
Cancers, 11(3) (2019-03-22)
Hepatocellular carcinoma (HCC) is one of the most prevalent and deadly cancers worldwide. Therefore, current global research focuses on molecular tools for early diagnosis of HCC, which can lead to effective treatment at an early stage. Perilipin 5 (PLIN5) has
Yu-Ting Yen et al.
Communications biology, 4(1), 658-658 (2021-06-04)
It has not been well studied which cells and related mechanisms contribute to endochondral ossification. Here, we fate mapped the leptin receptor-expressing (LepR+) mesenchymal stem cells (MSCs) in different embryonic and adult extremities using Lepr-cre; tdTomato mice and investigated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service