Skip to Content
Merck

Skip To

ABS487

Anti-LARS Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Leucine-tRNA ligase, cytoplasmic, Leucyl-tRNA synthetase, LeuRS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 348,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-

€ 348,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LARS(51520)

General description

LARS is the critical cytoplasmic enzyme that charges the tRNA for leucine to become L-leucyl-tRNA, or tRNA(Leu). LARS is thus critical for protein biosynthesis and LARS is up regulated, like many proteins, in cancer. LARS is localized in the cytoplasm and regulated via acetylation. Interestingly, recent research demonstrates that LARS and other tRNA synthetases play key roles as amino acid cellular sensors in my cellular signaling pathways such as mTOR which regulate cell size, protein translation, and even autophagy.
~140 kDa observed

Immunogen

KLH-conjugated linear peptide corresponding to human LARS near the C-terminus.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected LARS in human kidney and human liver tissue.
This Anti-LARS Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of LARS.

Analysis Note

Evaluated by Western Blotting in HUVEC cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected LARS in 10 µg of HUVEC cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
WH0023395M3HPA035951HPA045450
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice

shipped in

wet ice

clone

polyclonal

clone

3F12, monoclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters