Skip to Content
Merck
All Photos(1)

Key Documents

ABS482

Sigma-Aldrich

Anti-Perilipin-3, C-terminus Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Perilipin-3, Cargo selection protein TIP47, Mannose-6-phosphate receptor-binding protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€381.00

€381.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€381.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€381.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Plin3(66905)

General description

Perilipin-3, also known as Cargo selection protein TIP47, or Placental protein 17, PP17, 47 kDa mannose 6-phosphate receptor-binding protein, 47 kDa MPR-binding protein, also known as Mannose-6-phosphate receptor-binding protein 1, and encoded by the gene PLIN3/M6prbp1/Tip47, is a peripheral membrane protein that is required for the transport of mannose 6-phosphate receptors (MPR) from the endosomes to the trans-Golgi network. Perilipin 3 is part of the Perilipin family of proteins which are proteins that decorate the surface of cytosolic lipid droplets and play important roles in lipid metabolism. Cytosolic lipid droplets are now recognized as multifunctional organelles that, among other things, can accumulate in leukocytes under various inflammatory conditions. Perilipin-3 seems to play a critical role in the formation of lipid droplets and production of the the prostaglandin PGE2 an important inflammatory compound.

Immunogen

Epitope: C-terminus
Linear peptide corresponding to the C-terminus of mouse Perilipin-3, C-terminus.

Application

Immunocytochemistry Analysis: A representative lot detected Perilipin-3, C-terminus in OP9 cells (Skinner, J. R., et al. (2009). J Biol Chem. 284(45):30941-30948).
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-Perilipin-3 antibody, C-terminus is validated for use in western blotting & ICC for the detection of Perilipin-3.

Quality

Evaluated by Western Blotting in NIH/3T3-L1 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Perilipin-3, C-terminus in 10 µg of NIH/3T3-L1 cell lysate.

Target description

~ 52 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS without preservatives with 10% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie Hiel et al.
Nutrients, 10(5) (2018-04-26)
Postprandial hyperlipidemia is an important risk factor for cardiovascular diseases in the context of obesity. Inulin is a non-digestible carbohydrate, known for its beneficial properties in metabolic disorders. We investigated the impact of inulin on postprandial hypertriglyceridemia and on lipid
Amin Khalifeh-Soltani et al.
JCI insight, 1(18), e87418-e87418 (2016-11-05)
The small intestine has an underappreciated role as a lipid storage organ. Under conditions of high dietary fat intake, enterocytes can minimize the extent of postprandial lipemia by storing newly absorbed dietary fat in cytoplasmic lipid droplets. Lipid droplets can
Mika Yamada et al.
Journal of lipid research, 63(5), 100194-100194 (2022-03-15)
Lipid droplets (LDs) are multifunctional organelles that regulate energy storage and cellular homeostasis. The first step of triacylglycerol hydrolysis in LDs is catalyzed by adipose triglyceride lipase (ATGL), deficiency of which results in lethal cardiac steatosis. Although hormone-sensitive lipase (HSL)
Christophe A C Freyre et al.
Molecular cell, 76(5), 811-825 (2019-10-20)
Physical contact between organelles is vital to the function of eukaryotic cells. Lipid droplets (LDs) are dynamic organelles specialized in lipid storage that interact physically with mitochondria in several cell types. The mechanisms coupling these organelles are, however, poorly understood

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service