Skip to Content
Merck
All Photos(1)

Key Documents

ABS233-AF488

Sigma-Aldrich

Anti-PI3 Kinase p85 Antibody, N-SH2 domain Antibody, Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

Phosphatidylinositol 3-kinase regulatory subunit alpha, PI3-kinase regulatory subunit alpha, PI3K regulatory subunit alpha, PtdIns-3-kinase regulatory subunit alpha, Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha, PI3-kinase subunit p85-al

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

bovine ... Pik3R1(282307)
human ... PIK3R1(5295)
mouse ... Pik3R1(18708)
rat ... Pik3R1(25513)
rhesus monkey ... Pik3R1(698996)

General description

Phosphatidylinositol 3-kinase regulatory subunit alpha (UniProt: Q63787; also known as PI3-kinase regulatory subunit alpha; PI3K regulatory subunit alpha; PtdIns-3-kinase regulatory subunit alpha; Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha; PI3-kinase subunit p85-alpha; PtdIns-3-kinase regulatory subunit p85-alpha) is encoded by the Pik3r1 (Gene ID 25513) gene in rat. PI3 Kinase is a heterodimeric complex composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. It binds to activated (phosphorylated) protein tyrosine kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. PI3 Kinase catalyzes the phosphorylation of PI(4,5)P2 (PIP2) inositol ring at the 3 position to generate PI(3,4,5)P3 (PIP3), a potent second messenger that mediates the action of several growth factors. PI3 kinase is essential for the insulin signaling and also plays an important role in signaling in response to FGFR1, FGFR2, FGFR3, FGFR4, KITLG/SCF, KIT, PDGFRA and PDGFRB. Tyrosine phosphorylation of growth factor receptors creates docking sites for p85 binding (through its SH2 domains), bringing p85-associated p110 proximal to its lipid substrate on the membrane. PI3 Kinase is also activated by Ras and by the beta and gamma subunits of heterotrimeric G-proteins.

Specificity

This polyclonal antibody targets the first SH2 domain region common to human (isoform 1, AS53, p46, p85I), mouse, and rat (isoform p85-alpha, p55-alpha, and p50-alpha) spliced isoforms of PI3 kinase regulatory subunit alpha reported by UniProt (P27986, P26450, Q63787).

Immunogen

GST-tagged recombinant fragment corresponding to the first SH2 domain of rat PI3 kinase regulatory subunit alpha.

Application

Anti-PI3 Kinase p85, N-SH2 domain, Alexa Fluor 488 Conjugate, Cat. No. ABS233-AF488, detects PI3 Kinase p85 and is validated for use in immunocytochemistry.
Research Category
Signaling
The unconjugated antibody (Cat. No. ABS233) is also available for immunoprecipitation and Western blotting applications.

Quality

Evaluated by Immunocytochemistry in HepG2 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PI3 kinase p85 subunit in 4% paraformaldehyde-fixed, 0.1% Triton X-100-permeabilized HepG2 cells.

Target description

~85/50 kDa observed. 83.47/53.49/49.97/84.47 kDa (human isoform 1/AS53/p46/p85I; UniProt P27986), 83.39 kDa (mouse; UniProt P26450), 83.40/53.39/50.00 kDa (rat p85-alpha/p55-alpha/p50-alpha isoforms; UniProt Q63787).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody Alexa Fluor 488 conjugate in PBS with 1.5% BSA and 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service