Skip to Content
Merck

Skip To

ABS222

Anti-DEPTOR Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

DEP domain-containing mTOR-interacting protein, DEP domain-containing protein 6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
9
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DEPTOR(64798)

General description

DEPTOR (DEPDC6, DEP domain-containing protein 6) was recently identified as a novel mTOR-interacting protein whose over-expression in certain cell types represents a new mechanism for activating PI3K/Akt signaling and promoting cell survival. DEPTOR negatively regulates mTORC1 and mTORC2. It has been shown that loss of DEPTOR activates various targets that include p70S6K1, Akt, and SGK1, that leads to cell growth and survival. Little was known about DEPDC6 prior to the discovery of its interactions with mTOR, at which time it was given the new name of DEPTOR (Peterson, T.R., et al. (2009). Cell. 137(5):873-876).
~46 kDa observed.

Immunogen

Epitope: DEP domain-containing mTOR-interacting protein DEPTOR.
KLH-conjugated linear peptide corresponding to the DEP domain-containing mTOR-interacting protein of human DEPTOR.

Application

Anti-DEPTOR Antibody detects level of DEPTOR & has been published & validated for use in WB.
Research Category
Metabolism
Research Sub Category
PI3K, Akt, & mTOR Signaling

Biochem/physiol Actions

Other homologies: Primate, Equine, Porcine, Canine, Rat, and Mouse based on 99%, 99%, 98%, 97%, and 96% homology, respectively.
This antibody recognizes the DEP domain that contains mTOR-interacting protein DEPTOR.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lyaste
Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected DEPTOR in HEK293 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 09-463

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA023945HPA024011ABS79
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Gene Information

human ... DEPTOR(64798)

Gene Information

human ... DEPDC6(64798)

Gene Information

human ... DEPDC6(64798)

Gene Information

human ... MTOR(2475)

UniProt accession no.

Q8TB45

UniProt accession no.

-

UniProt accession no.

-

UniProt accession no.

P42345


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters