Skip to Content
Merck
All Photos(2)

Documents

ABS194

Sigma-Aldrich

Anti-phospho PDHE1-A type I (Ser300) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial, PDHE1-A type I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology), bovine (based on 100% sequence homology), Xenopus (based on 100% sequence homology), zebrafish (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer300)

Gene Information

human ... PDHA1(5160)

General description

In many organisms, the pyruvate dehydrogenase complex catalyzes the overall, irreversible conversion of pyruvate to acetyl-CoA and CO2 in the aerobic, energy-generating pathways, in addition to serving as a key regulator for cardiac substrate selection. It contains multiple copies of three enzymatic components: pyruvate dehydrogenase (E1), dihydrolipoamide acetyltransferase (E2) and lipoamide dehydrogenase (E3). PDH is regulated by both pyruvate dehydrogenase kinase (PDK)-mediated phosphorylation and feedback inhibition.

Specificity

This antibody recognizes PDHE1-A type I phosphorylated at Ser300.

Immunogen

Epitope: Phosphorylated Ser300
KLH-conjugated linear peptide corresponding to human PDHE1-A type I phosphorylated at Ser300.

Application

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected PDHE1-A type I in dichloroacetate untreated and treated HEK 293 cells.
This Anti-phospho PDHE1-A type I (Ser300) Antibody is validated for use in WB, IC for the detection of phospho PDHE1-A type I (Ser300).

Quality

Evaluated by Western Blot in dichloroacetate untreated and treated HEK 293 cell lysates.

Western Blot Analysis: 0.5 µg/mL of this antibody detected PDHE1-A type I on 10 µg of dichloroacetate untreated and treated HEK 293 cell lysates.

Target description

~43 kDa observed

Analysis Note

Control
Dichloroacetate untreated and treated HEK 293 cell lysates

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinxin Liu et al.
Theranostics, 11(17), 8283-8300 (2021-08-11)
Rationale: The molecular mechanisms underlying the pathogenesis of systemic insulin resistance in type 2 diabetes remain elusive. Growth hormone receptor (GHR) deficiency has long been known to improved insulin sensitivity. However, whether hepatic GHR overexpression or activation is a cause
Yuan Z Feng et al.
Journal of lipid research, 58(11), 2147-2161 (2017-08-22)
Lipid droplet (LD) coating proteins are essential for the formation and stability of intracellular LDs. Plin2 is an abundant LD coating protein in skeletal muscle, but its importance for muscle function is unclear. We show that myotubes established from Plin2-/-
Anders Kalsen et al.
Journal of applied physiology (Bethesda, Md. : 1985), 117(10), 1180-1187 (2014-09-27)
In a randomized, double-blind crossover design, we investigated the effect of the beta2-agonist terbutaline (TER) on endurance performance and substrate utilization in nine moderately trained men [maximum oxygen uptake (V̇O(2 max)) 58.9 ± 3.1 ml·min(-1)·kg(-1)]. Subjects performed 60 min of
G Perez-Siles et al.
Scientific reports, 10(1), 9262-9262 (2020-06-07)
Charcot-Marie-Tooth (CMT) is a group of inherited diseases clinically and genetically heterogenous, characterised by length dependent degeneration of axons of the peripheral nervous system. A missense mutation (p.R158H) in the pyruvate dehydrogenase kinase 3 gene (PDK3) has been identified as
Kenichi Aizawa et al.
Cells, 13(1) (2024-01-11)
Heart failure with reduced ejection fraction (HFrEF) is characterized not only by reduced left ventricular ejection fraction (EF) but is also combined with symptoms such as dyspnea, fatigue, and edema. Several pharmacological interventions have been established. However, a treatment targeting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service