Skip to Content
Merck
All Photos(1)

Documents

ABS1641

Sigma-Aldrich

Anti-SPTLC2 Antibody

serum, from rabbit

Synonym(s):

Serine palmitoyltransferase 2, hLCB2a, LCB 2, LCB2a, Long chain base biosynthesis protein 2, Long chain base biosynthesis protein 2a, Serine-palmitoyl-CoA transferase 2, SPT 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

chicken (based on 100% sequence homology), rat (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), Xenopus (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SPTLC2(9517)

General description

Serine palmitoyltransferase 2 (EC 2.3.1.50; UniProt O15270; also known as hLCB2a, LCB 2, LCB2a, Long chain base biosynthesis protein 2, Long chain base biosynthesis protein 2a, Serine-palmitoyl-CoA transferase 2, SPT 2) is encoded by the SPTLC2 (also known as HSAN1C, KIAA0526, NSAN1C, LCB2) gene (Gene ID 9517) in human. Serine palmitoyltransferase (SPT) is a pyridoxal 5′-phosphate (PLP)-dependent enzyme that catalyses the first step of de novo sphingolipids (SLs) biosynthesis, namely the condensation of L-serine with palmitoyl coenzyme-A to generate 3-ketodehydrosphinganine (KDS) that forms the sphingoid base backbone of all SLs. The core human SPT homoenzyme is a membrane-bound heterodimer composed of an hLCB1 (SPTLC1) subunit together with either an hLCB2a (SPTLC2) or an hLCB2b (SPTLC3) subunit. The hLCB1/hLCB2 heterodimer contains a single active site composed of residues from both subunits. hLCB2a and hLCB2b share 68% sequence identity, with hLCB2b being the isotype that is predominantly expressed in the placenta and prefers shorter chain acyl-CoAs (C12 and C14) to generate short-chain SLs. In addition, two human small subunits (ssSPTa and ssSPTb) have been identified and found to maximize the catalytic activity and to confer the acyl-CoA substrate chain-length specificity of the hLCB1/hLCB2 core enzyme. Mutations in hLCB1 and hLCB2a are found in patients with hereditary sensory and autonomic neuropathy type I (HSAN1), an inherited disorder that affects sensory and autonomic neurons due to neurotoxic deoxysphingolipids formation as a result of substrate promiscuity of the mutated SPT enzyme.

Specificity

Target-specificity of this antiserum was verified by negative blotting using pre-immune serum and by target band blocking using immunogen peptides (Hornemann, T., et al. (2006). J. Biol. Chem. 281(49):37275-37281).

Immunogen

A mixture of two linear peptides corresponding to sequences from the N-terminal region and the C-terminal end of human SPTLC2.
Epitope: N-terminal region and C-terminal end.

Application

Anti-SPTLC2 Antibody is an antibody against SPTLC2 for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Lipid Signaling
Western Blotting Analysis: Representative lots detected SPTLC2 in human placenta tissue homogenates and in various lysates from human cell lines, including embryonic kidney HEK293, hepatocellular carcinoma HepG2, choriocarcinoma JAr and JEG-3 (Hornemann, T., et al. (2007). Biochem. J. 405(1):157-164; Hornemann, T., et al. (2006). J. Biol. Chem. 281(49):37275-37281).

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected SPTLC2 in 10 µg of HepG2 cell lysate.

Target description

~63 kDa observed. 62.92 kDa calculated. Target band size is also reported with a smaller apparent molecular weight of ~58kDa Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit polyclonal antibody serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deanna L Davis et al.
Journal of lipid research, 61(4), 505-522 (2020-02-12)
Myelin is a unique lipid-rich membrane structure that accelerates neurotransmission and supports neuronal function. Sphingolipids are critical myelin components. Yet sphingolipid content and synthesis have not been well characterized in oligodendrocytes, the myelin-producing cells of the CNS. Here, using quantitative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service