Skip to Content
Merck
All Photos(3)

Key Documents

ABS1494

Sigma-Aldrich

Anti-SCYL1

from rabbit, purified by affinity chromatography

Synonym(s):

N-terminal kinase-like protein, Coated vesicle-associated kinase of 90 kDa, SCY1-like protein 1, Telomerase regulation-associated protein, Telomerase transcriptional element-interacting factor, Teratoma-associated tyrosine kinase

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€131.00
100 μG
€379.00

€131.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€131.00
100 μG
€379.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€131.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SCYL1(57410)

General description

N-terminal kinase-like protein (UniProt: Q96KG9; also known as Coated vesicle-associated kinase of 90 kDa, SCY1-like protein 1, Telomerase regulation-associated protein, Telomerase transcriptional element-interacting factor, Teratoma-associated tyrosine kinase) is encoded by the SCYL1 (also known as CVAK90, GKLP, NTKL, TAPK, TEIF, TRAP, HT019) gene (Gene ID: 57410) in human. SCY1-like protein 1 is ubiquitously expressed and it regulates COPI-mediated retrograde protein traffic at the interface between the Golgi apparatus and the endoplasmic reticulum. It is involved in the maintenance of the Golgi apparatus morphology. It has a divergent N-terminal kinase domain (aa 14-314), but it is catalytically inactive. It also contain three HEAT repeats (aa 350-388; 389-427; and 507-545). Six isoforms of SCY1-like protein 1 have been reported that are produced by alternative splicing. Its intracellular distribution is isoform specific. Isoforms 1 and 2 are shown to be cytoplasmic throughout the cell cycle. Isoform 3 is cytoplasmic during interphase and centrosomal during mitosis. Isoform six is generally localized to nucleus and it acts as a transcriptional activator and binds to three different types of GC-rich DNA binding sites (box-A, -B and -C) in the beta-polymerase promoter region. Mutations in SCYL1 gene are reported to cause Spinocerebellar ataxia, a clinically and genetically heterogeneous group of cerebellar disorders due to degeneration of the cerebellum with variable involvement of the brainstem and spinal cord.

Specificity

This rabbit polyclonal antibody detects SCY1-Like Protein 1 (SCYL1) in human cells. It targets an epitope within 16 amino acids from the C-terminal region. The immunogen is conserved between isoforms 1, 2, and 3.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human SCY1-Like Protein 1.

Application

Anti-SCYL1, Cat. No. ABS1494, is a rabbit polyclonal antibody that detects SCY1-Like Protein 1 and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SCYL1 in HeLa, A431, HUVEC, and NIH/3T3 cells.
Research Category
Signaling

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected SCY1-Like Protein 1 (SCYL1) in HeLa cell lysate.

Target description

~100 kDa observed; 89.63 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service