Skip to Content
Merck
All Photos(2)

Key Documents

ABN96

Sigma-Aldrich

Anti-Neurexin3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Neurexin-3-alpha, Neurexin III-alpha

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€385.00

€385.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€385.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€385.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NRXN3(9369)

General description

Neurexins are heavily glycosylated and processed neuronal transmembrane cell surface proteins likely involved in cell recognition and adhesion. (Missler, M., et al. (1998) 14(1):20-26) Antibody Anti-neurexin 3 alpha (ABN96) was made against a peptide immunogen corresponding to a stretch of amino acids in the extracellular domain of human neurexin 3 alpha (Uniprot Q9Y4C0). Although the theoretical full length of Neurexin 1alpha is 180 kDa, the protein detected in human brain lysates may show a band around 180 kDa and additional bands around 40-55 kDa as a possible result of the posttranslational processing previously reported.

Immunogen

KLH-conjugated linear peptide corresponding to human Neurexin-3.

Application

Detect Neurexin3 using this Anti-Neurexin3 Antibody validated for use in WB, IH.
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected Neurexin-3 in normal mouse brain tissue.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blot in PC12 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Neurexin-3 in 10 µg of PC12 cell lysate.

Target description

~180 kDa observed. UniProtKB/Swiss-Prot Q9Y4C0 (NRX3A_HUMAN) states 2 isoforms due to alternative promoter usage and alternative splicing at a molecular weight of ~181 kDa and ~117 kDa, with several beta isoforms at a molecular weight 70 kDa, 51 kDa, 47 kDa, and 44 kDa Additional bands may also be observed around 40-55 kDa as a possible result of the posttranslational processing.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
PC12 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongmei Yang et al.
Science advances, 8(50), eade0764-eade0764 (2022-12-15)
Protein S-nitros(yl)ation (SNO) is a posttranslational modification involved in diverse processes in health and disease and can contribute to synaptic damage in Alzheimer's disease (AD). To identify SNO proteins in AD brains, we used triaryl phosphine (SNOTRAP) combined with mass

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service