Skip to Content
Merck
All Photos(3)

Key Documents

ABN464

Sigma-Aldrich

Anti-IRE1p Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Endoplasmic reticulum-to-nucleus signaling 1, Serine/threonine-protein kinase/endoribonuclease IRE1, ERN1, IRE1, Inositol-requiring protein 1, hIRE1p, Ire1-alpha, IRE1a

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€379.00

€379.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€379.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€379.00


Estimated to ship on16 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ERN1(2081)

General description

Inositol-requiring protein 1 (IRE1p), also known as Ire1-alpha (IRE1a), Endoplasmic reticulum-to-nucleus signaling 1 (ERN1), and Serine/threonine-protein kinase/endoribonuclease IRE1, contains 1 KEN domain and 1 protein kinase domain.
IRE1p has an endoribonuclease activity that is required to splice X-box binding protein (XBP1) mRNA converting it to a potent UPR transcriptional activation. Depletion of IRE1p through the expression of a dominant negative form of IRE1p has no effect on transfected cells, but cell death via apoptosis occurs under stress conditions that cause unfolded proteins to accumulate in the ER. IRE1p is ubiquitously expressed with high levels observed in pancreatic tissue.

Specificity

This antibody recognizes the C-terminus of IRE1p.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human IRE1p.

Application

Detect Inositol-requiring protein 1 using this rabbit polyclonal antibody, Anti-IRE1p Antibody validated for use in western blotting, ICC & IHC.
Immunohistchemistry Analysis: A 1:1,000 dilution from a representative lot detected IRE1p in human pancreas and kidney tissues.

Immunocytochemistry Analysis: 1 µg/mL from a representative lot detected IRE1p in A20 cells.

Immunofluorescence Analysis: 2 µg/mL from a representative lot detected IRE1p in A20 cells.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blotting in A20 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected IRE1p in A20 cell lysate.

Target description

~114 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A20 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service