Skip to Content
Merck
All Photos(3)

Documents

ABN415

Sigma-Aldrich

Anti-TRAPPC1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

TRAPPC-1, Trafficking protein particle complex subunit 1, BET5 homolog, Multiple myeloma protein 2, MUM-2, MUM2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TRAPPC1(58485)

Related Categories

General description

TRAPPC1 is one part of a larger protein complex called the TRAPP (transport protein particle) complex vital to Golgi function and transport. TRAPPC1 is localized to the Golgi and endoplasmic reticulum. Vesicle-mediated transport is a process carried out by virtually all cells and is required for the proper targeting and secretion of proteins. As such, there are numerous proteins involved to ensure that the proteins are properly transported, targeted, tethered, and localized. The TRAPP complex is just one such complex but it is one of the most studied. Mutations in TRAPPC proteins can lead to serious disorders including muscular dystrophy and myopathy such as that seen with mutations associated with TRAPPC11 (a related gene to TRAPPC1).

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to the Near C-terminus of Human TRAPPC1.

Application

Research Category
Neuroscience

Neuroscience
Research Sub Category
Ion Channels & Transporters

Neurotransmitters & Receptors
This Anti-TRAPPC1 Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of TRAPPC1.
Western Blotting Analysis: 0.25 µg/mL from a representative lot detected TRAPPC1 in 5 µg of mouse brain tissue lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected TRAPPC1 in human cerebral cortex, cerebellum, and kidney.

Quality

Evaluated by Western Blotting in TRAPPC1 transfected HEK293T cell lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected TRAPPC1 in 5 µg of TRAPPC1 transfected HEK293T cell lysate.

Target description

~19 kDa observed. The endogenous band appears at ~31kDa (mouse brain) while the transfected band appears at ~19kDa

Physical form

Antigen Affinity Purified
Purified rabbit Polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service