Skip to Content
Merck
All Photos(1)

Key Documents

ABN368

Sigma-Aldrich

Anti-Spastin Antibody

serum, from rabbit

Synonym(s):

Spastin, Spastic paraplegia 4 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€435.00

€435.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€435.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€435.00


Estimated to ship on01 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SPAST(6683)

General description

Spastin is a distinct microtubule-severing protein. Autosomal dominant mutations in the SPG4 locus corresponding to spastin are the most common cause of hereditary spastic paraplegia (HSP), a neurodegenerative disease that afflicts the adult corticospinal tracts.

Immunogen

Recombinant protein corresponding to human Spastin with 4 isoforms.

Application

Detect Spastin using this rabbit polyclonal antibody, Anti-Spastin Antibody validated for use in western blotting & ICC.
Western Blotting Analysis: A representative lot from an independent laboratory detected Spastin in myc-Spastin transfected HeLa and MRC5 cell lysates (Connell, et al. (2009). Traffic. 10:42-56).

Quality

Evaluated by Western Blotting in L6 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Spastin in 10 µg of L6 cell lysate.

Target description

~ 68, 64, 58, 55 kDa observed. Uniprot describes 4 isoforms produced by alternative splicing at ~68 kDa, ~64 kDa, ~58 kDa, and ~55 kDa Spastin has shown in some western blots that four isoforms [full length (68 kD), exon 4-deleted full length (64 kD), short form (58-60 kD) and exon 4-deleted short form (55 kD)] are expressed in mammalian cells. Ths pAb detects bands corresponding to the size of these four isoforms in HeLa, MRC5 human lung fibroblast and NSC34 mouse lower motor neuron neuroblastoma fusion cell lines (Connell, et al. (2009). Traffic. 10:42-56).

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiuling Liu et al.
eLife, 12 (2024-01-17)
Axon regeneration is abortive in the central nervous system following injury. Orchestrating microtubule dynamics has emerged as a promising approach to improve axonal regeneration. The microtubule severing enzyme spastin is essential for axonal development and regeneration through remodeling of microtubule
Chi-Lun Chang et al.
The Journal of cell biology, 218(8), 2583-2599 (2019-06-23)
Lipid droplets (LDs) are neutral lipid storage organelles that transfer lipids to various organelles including peroxisomes. Here, we show that the hereditary spastic paraplegia protein M1 Spastin, a membrane-bound AAA ATPase found on LDs, coordinates fatty acid (FA) trafficking from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service