Skip to Content
Merck
All Photos(3)

Key Documents

ABN333

Sigma-Aldrich

Anti-Neurotrypsin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Neurotrypsin, Leydin, Motopsin, Serine protease 12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Neurotrypsin is a central nervous system-expressed serine protease whose truncation or absence causes nonsyndromic mental retardation. It is most prominently expressed in structures that are involved in the processing and storage of learned behavior and memory, such as the cerebral cortex, the hippocampus, and amygdala. Evidence suggests that neurotrypsin has multiple functions. These functions include axonal outgrowth, maintaining neuronal plasticity, and arranging the perineuronal environment, partly in coordination with other proteases including tissue plasminogen activator. At least two isoforms of neurotrypsin are known to exist.

Immunogen

KLH-conjugated linear peptide corresponding to human Neurotrypsin.

Application

Detect the Neurotrypsin protein using this Anti-Neurotrypsin validated for use in WB & IHC.
Western Blot Analysis: 1 µg/mL from a representative lot detected Neurotrypsin in 10 µg/mL of rat testis tissue lysate.

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Neurotrypsin in human midbrain and rat cerebellum tissues.

Quality

Evaluated by Western Blot in mouse testis tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Neurotrypsin in 10 µg/mL of mouse testis tissue lysate.

Target description

~97 kDa observed. Uncharacterized bands may appear at ~13 kDa, ~38 kDa, and ~43 kDa in some lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service