Skip to Content
Merck
All Photos(2)

Key Documents

ABN309

Sigma-Aldrich

Anti-SAPAP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

SAP90, PSD-95-associated protein 1, Disks large-associated protein 1, DLGAP1, DAP-1, GKAP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€335.00

€335.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€335.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€335.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DLGAP1(9229)

General description

PSD-95/SAP90-binding protein 1 (SAPAP1) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP1 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). SAPAP1 mRNA is targeted to cell bodies in a similar manner to SAPAP2 and -4, whereas SAPAP3 mRNA is detected mainly in cell bodies. SAPAP1 protein however, is targeted to the synapse and is not reliant on the synaptic localization of PSD-95 or the synaptic scaffolding molecule (S-SCAM).

Specificity

At least three isoforms of SAPAP1 are known to exist. This SAPAP1 antibody will not cross-react with other SAPAP proteins.

Immunogen

KLH-conjugated linear peptide corresponding to human SAPAP1.

Application

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected SAPAP1 in human brain tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This SAPAP1 antibody is validated for use in WB & IHC for the detection of the SAPAP1 protein.

Quality

Evaluated by Western Blot in human brain tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected SAPAP1 in 10 µg of human brain tissue lysate.

Target description

~95 kDa observed. Uniprot describes 7 isoforms ranging between ~70 and ~109 kDa An uncharacterized band may appear at ~58 kDa in some lysates. The calculated molecular weight is 109 kDa, however SAPAP1 has been shown as a ~95 kDa band in western blots. (Naisbitt, S., et al. (1997). 17(15):5687-5696.)

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service