Skip to Content
Merck
All Photos(2)

Key Documents

ABN257

Sigma-Aldrich

Anti-NSG1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Neuron-specific protein family member 1, Brain neuron cytoplasmic protein 1, NSG1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€326.00

€326.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€326.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€326.00


Estimated to ship on27 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NSG1(27065)

General description

Neuron-specific protein family member 1 (NSG1), is also known as Brain neuron cytoplasmic protein 1. It is responsible for dopamine receptor binding in the dopamine signaling pathway.

Specificity

This antibody recognizes the cytoplasmic domain of NSG1.

Immunogen

Epitope: Cytoplasmic domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human NSG1.

Application

Detect the NSG1 protein using this Anti-NSG1 validated for use in WB & IHC.
Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected NSG1 in rat cerebellum, human pons/midbrain, and rat hippocampus tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in human cerebellum tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected NSG1 in 10 µg of human cerebellum tissue lysate.

Target description

~20/17 kDa observed. An uncharacterized band may appear at ~12 kDa in some lysates. The calculated molecular weight is 21 kDa, however NSG1 has been shown as a ~20/17 kDa band in western blots (Catsicas, S., et al. (2004). Lausanne, EPFL. 1-162).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human cerebellum tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service