Skip to Content
Merck
All Photos(1)

Key Documents

ABN254

Sigma-Aldrich

Anti-BAALC Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Brain and acute leukemia cytoplasmic protein, BAALC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology), bovine (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... BAALC(79870)

General description

Brain and acute leukemia cytoplasmic protein (BAALC) interacts with CAMK2A in a synaptic role at postsynaptic lipid rafts. BAALC is expressed by neural, hematopoietic, normal early progenitor cells and in the most immature type of blasts in acute leukemia. Isoform 2 and isoform 6 are also expressed in the brain.

Specificity

This antibody recognizes the CAMK2A interaction domain of BAALC. This antibody should recognize all isoforms in uniprot.

Immunogen

Epitope: CAMK2A interaction domain
KLH-conjugated linear peptide corresponding to the CAMK2A interaction domain of human BAALC.

Application

Detect Brain & acute leukemia cytoplasmic protein using this rabbit polyclonal antibody, Anti-BAALC Antibody validated for use in western blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected BAALC in 10 µg of mouse brain tissue lysate.

Target description

~22 kDa observed. Uniprot describes 6 isoforms produced by alternative splicing at ~19 kDa, ~16 kDa, ~9 kDa, ~8 kDa and ~6 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service