Skip to Content
Merck
All Photos(3)

Key Documents

ABN2242

Sigma-Aldrich

Anti-Vangl2

from rabbit, purified by affinity chromatography

Synonym(s):

Vang-like protein 2 (UniProt: Q9ULK5, also known as Loop-tail protein 1 homolog, Strabismus 1, Van Gogh-like protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€154.00
100 μG
€403.00

€154.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€154.00
100 μG
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€154.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... VANGL2(57216)

General description

Vang-like protein 2 (UniProt: Q9ULK5; also known as Loop-tail protein 1 homolog, Strabismus 1, Van Gogh-like protein 2) is encoded by the VANGL2 (also known as KIAA1215, STB1) gene (Gene ID: 57216) in human. Vangl2 is a homodimeric multi-pass membrane protein that plays a role in the control of early morphogenesis, patterning of both axial midline structures and the development of neural plate. It can also form heterodimer with VANGL1. It is required for polarization and movement of myocardializing cells in the outflow tract and seems to act via RHOA signaling to regulate this process. Vang-like protein 2 is reported to bind to Dvl to cell-autonomously promote efficient Dvl plasma membrane recruitment, a pre-requisite for planar cell polarity activation. At the same time, it can inhibit Dvl from interacting with its downstream effector (Dishevelled associated activator of morphogenesis 1 (Daam1), and functionally suppresses Dvl - Daam1 cascade. Mutations in VANGL2 gene cause neural tube defects that results in defective neural tube closure during the first trimester of pregnancy. (Ref.: Seo, HS et al. (2017). Hum. Mol. Genet. 26(11); 2053-2061).

Specificity

This rabbit polyclonal antibody detects Vang-like protein 2 in human and murine cells. It targets an epitope with in 15 amino acids from the N-terminal region.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal region of human Vang-like Protein 2.

Application

Anti-Vangl2, Cat. No. ABN2242, is a highly specific rabbit polyclonal antibody that targets Vang-like protein 2 and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Vangl2 in HT1080 cells.

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Vangl2 in human tonsil and mouse embryonic spinal cord tissue sections.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Vangl2 in HepG2 cell lysate.

Target description

~60 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service