Skip to Content
Merck
All Photos(1)

Key Documents

ABN201

Sigma-Aldrich

Anti-CYP24A1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

1,25-dihydroxyvitamin D(3) 24-hydroxylase, mitochondrial, 24-OHase, Vitamin D(3) 24-hydroxylase, Cytochrome P450 24A1, Cytochrome P450-CC24

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€326.00

€326.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€326.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€326.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CYP24A1(1591)

General description

Also known as Vitamin D(3) 24-hydroxylase, CYP24A1 is a member of the cytochrome P450 (CYP) family of enzymes. Most members of the CYP family can metabolize multiple substrates, and can catalyze multiple reactions. The CYP24A1 is a mitochondrial protein that starts the breakdown of 1,25-dihydrozyvitamin D3 by hydroxylation of the side chain to form calcitroic acid. This enzyme also has a role in calcium homeostasis and the vitamin D endocrine system.

Specificity

Other Homologies: Bovine (92% sequence homology) and Rat (75% sequence homology).

Immunogen

KLH-conjugated linear peptide corresponding to human CYP24A1.

Application

Anti-CYP24A1 Antibody is an antibody against CYP24A1 for use in 0.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blot in A549 cell lysate.

Western Blot Analysis: 0.2 µg/mL of this antibody detected CYP24A1 in 10 µg of A549 cell lysate.

Target description

~60 kDa observed. Two isoforms are known to exist at 59 kDa and 51 kDa This antibody is expected to recognize both isoforms.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A549 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service