Skip to Content
Merck
All Photos(2)

Documents

ABN1000

Sigma-Aldrich

Anti-MAGL Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Monoglyceride lipase, MGL, Monoacylglycerol lipase, MAGL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MGLL(11343)

General description

Monoglyceride lipase (MGL), or alternatively HU-K5, Lysophospholipase homolog, Lysophospholipase-like, or Monoacylglycerol lipase (MAGL) is a protein encoded by the MGLL gene in humans and is very important in lipid metabolism. Monoglyceride lipase is the enzyme that converts monoacylglycerides (key building blocks of lipids) into free fatty acid chains and glycerol. Also, Monoglyceride Lipase hydrolyzes endocannabinoids which ultimately can regulate nociperception and the perception of pain, so the enzyme is being studied in pain mediation therapies. Monoglyceride Lipase is expressed in many tissues including fat, lung, liver, brain and heart. In disease, Monoglyceride Lipase is being studied most intensely in cancer research. In some cancers it appears to be play a suppressive role in regulating AKT mediated signaling, but in others, since the enzyme regulates the levels of fatty acids that can serve as intra and intercellular signaling molecules, Monoglyceride lipase activity seems to promote cancer cell migration, invasion and growth.

Immunogen

Recombinant protein corresponding to mouse MAGL.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected MAGL in human cerebral cortex tissue.
Immunohistochemistry Analysis: A representative lot detected MAGL in human hippocampus tissue (Mulder, J., et al. (2011). Brain. 134:1041-1060).
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-MAGL Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of MAGL.

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected MAGL in 10 µg of mouse brain tissue lysate.

Target description

~ 31/33 kDa observed. This protein can be alternatively spliced, so western blots may show a doublet. Evidence for alternative splicing of MAGL, can run as doublet, ~31 and ~33 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan D Shepard et al.
Frontiers in synaptic neuroscience, 13, 689518-689518 (2021-06-15)
Adverse events and childhood trauma increase the susceptibility towards developing psychiatric disorders (substance use disorder, anxiety, depression, etc.) in adulthood. Although there are treatment strategies that have utility in combating these psychiatric disorders, little attention is placed on how to
Ping-Yuan Wang et al.
Cancer prevention research (Philadelphia, Pa.), 14(1), 31-40 (2020-09-23)
Germline mutations of TP53, which cause the cancer predisposition disorder Li-Fraumeni syndrome (LFS), can increase mitochondrial activity as well as fatty acid β-oxidation (FAO) in mice. Increased fatty acid metabolism can promote cancer malignancy, but its specific contribution to tumorigenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service