Skip to Content
Merck
All Photos(1)

Key Documents

ABF292

Sigma-Aldrich

Anti-ZC3H12A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Ribonuclease ZC3H12A, MCP-induced protein 1, Zinc finger CCCH domain-containing protein 12A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€370.00

€370.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€370.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ZC3H12A(80149)

General description

Ribonuclease ZC3H12A, also known as MCP-induced protein 1 or Zinc finger CCCH domain-containing protein 12A, and encoded by the gene name ZC3H12A or MCPIP, is an essential member of a family of novel CCCH-zinc finger proteins that regulate macrophage activation. ZC3H12A may be involved in host immunity and inflammatory diseases. It has RNase activity that prevents some immune disorders by directly affecting the mRNA stability of interleukins such as IL-6 and IL12p40. Mice lacking the ZC3H12A gene suffered from severe anemia, and most dies within 12 weeks. Overexpression of ZC3H12A causes ER stress and induces a number of genes involved in apoptosis and autophagy, including JNK, PUMA, and beclin-1 in cardiac myoblasts, leading to cell death. ZC3H12A can also induce adipogenesis in 3T3-L1 pre-adipocytes in the absence of PPAPγ.

Immunogen

Epitope: center region
KLH-conjugated synthetic peptide corresponding to the center region of human ZC3H12A.

Application

Anti-ZC3H12A Antibody detects level of ZC3H12A & has been published & validated for use in ZC3H12A.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blotting in Raji Cell lysate.

Western Blotting Analysis: 2 μg/mL of this antibody detected ZC3H12A in Raji Cell lysate.

Target description

~75 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service