Skip to Content
Merck
All Photos(2)

Key Documents

ABF202

Sigma-Aldrich

Anti-IRF7 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Interferon regulatory factor 7, IRF7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€487.00

€487.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€487.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€487.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IRF7(3665)

General description

Interferon regulatory factor 7 (IRF7) is a member of the IRF family and has 1 IRF tryptophan pentad repeat DNA-binding domain. IRF7 is key transcriptional regulator of type I interferon (IFN)-dependent immune responses and regulates the transcription of type I IFN genes (IFN-alpha and IFN-beta) and IFN-stimulated genes (ISG) by binding to an interferon-stimulated response element (ISRE) in their promoters. IRF7 plays an important role in the innate immune response against DNA and RNA viruses and has been shown to activate both the IFN-beta (IFNB) and the IFN-alpha (IFNA) genes and mediate their induction via both the virus-activated, MyD88-independent pathway and the TLR-activated, MyD88-dependent pathway. IRF7 exists in an inactive form in the cytoplasm of uninfected cells and following viral infection it becomes phosphorylated by IKBKE and TBK1 kinases. This leads to its dimerization and nuclear localization where along with other coactivators it can activate transcription of the type I IFN and ISG genes. Studies show that IRF7 induces PSMB9/LMP2 expression, either directly or through induction of IRF1 and may play a role in regulating adaptive immune responses. Other evidence shows that IRF7 binds to the Q promoter (Qp) of EBV nuclear antigen 1 a (EBNA1) and may play a role in the regulation of EBV latency. Additionally, IRF7 is implied to activate distinct gene expression programs in macrophages and regulate the anti-tumor properties of primary macrophages. IRF7 is expressed predominantly in spleen, thymus and peripheral blood leukocytes.

Specificity

This antibody recognizes the C-terminus of IRF7.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human IRF7.

Application

Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected IRF7 in mouse spleen tissue.

Immunofluorescence Analysis: 20 µg/mL from a representative lot detected IRF7 in mouse spleen cells.
Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling
This Anti-IRF7 Antibody is validated for use in Western Blotting and Immunohistochemistry and Immunofluorescence for the detection of IRF7.

Quality

Evaluated by Western Blotting in HEK293 whole cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected IRF7 in HEK293 whole cell lysate.

Target description

~51 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 whole cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service