Skip to Content
Merck

Skip To

ABF199

Anti-TIM-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Hepatitis A virus cellular receptor 1, HAVcr-1, Kidney injury molecule 1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIMD-1, T-cell membrane protein 1, TIM-1, TIM, KIM1, TIM1, TIMD1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 358,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IHC, WB
Citations:
1

€ 358,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable, immunohistochemistry: suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HAVCR1(26762)

General description

T-cell immunoglobulin and mucin domain-containing protein 1 (TIMD-1, TIM-1, TIM, TIM1, TIMD1), also known as Hepatitis A virus cellular receptor 1 (HAVcr-1) and Kidney injury molecule 1(KIM-1, KIM1), may play an important role in T-helper cell development and the regulation of asthma and allergic diseases. In the case of human hepatitis A virus (HHAV) infection, TIM-1 functions as a cell-surface receptor for the virus, through which the hepatitis A virus can gain entry into a cell. Additional evidence implies that it may play a role in kidney injury and repair. TIM-1 is widely expressed, with highest levels in kidney and testis. It is also expressed by activated CD4+ T-cells during the development of helper T-cells responses. Furthermore, TIM-1 is expressed on T helper 2 (Th2) cells of the immune system, and following the binding of its natural ligand TIM-4, stimulates T cell expansion and cytokine production in response to viral challenge.
~48 kDa observed. Uncharacterized bands may appear in some lysate(s).

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human TIM-1.

Application

Anti-TIM-1 Antibody is a highly specific rabbit polyclonal antibody, that targets T-cell immunoglobulin & mucin domain-containing protein 1 & has been tested in western blotting, IHC & Immunofluorescence.
Immunohistochemistry Analysis: 10 µg/mL from a representative lot detected TIM-1 in human uterus tissue.

Immunofluorescence Analysis: 20 µg/mL from a representative lot detected TIM-1 in human uterus tissue.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Biochem/physiol Actions

This antibody recognizes the N-terminus of TIM-1.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human uterus tissue lysate
Evaluated by Western Blotting in human uterus tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected TIM-1 in 15 µg of human uterus tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB3500252MABF230MABF73
biological source

rabbit

biological source

rabbit

biological source

rat

biological source

mouse

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice

shipped in

wet ice

technique(s)

immunofluorescence: suitable, western blot: suitable, immunohistochemistry: suitable

technique(s)

immunofluorescence: suitable, immunohistochemistry: suitable, indirect ELISA: suitable, western blot: suitable

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable

technique(s)

flow cytometry: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters