Skip to Content
Merck
All Photos(2)

Key Documents

ABF1168

Sigma-Aldrich

Anti-Chitinase-1 (CHIT1)

from rabbit, purified by affinity chromatography

Synonym(s):

Chitotriosidase-1, EC: 3.2.1.14, Chitinase-1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€131.00
100 μL
€343.00

€131.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€131.00
100 μL
€343.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€131.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CHIT1(1118)

General description

Chitotriosidase-1 (UniProt: Q13231; also known as EC: 3.2.1.14, Chitinase-1) is encoded by the CHIT1 gene (Gene ID: 1118) in human. Chitinase-1 is a member of the Chitinase class II subfamily that degrades chitin, chitotriose and chitobiose. It is one of the most abundantly secreted protein and is mainly produced by activated macrophages and epithelial cells. Chitinase -1 can hydrolyze the beta-(1, 4)-linkage between the adjacent N-acetyl glucosamine residues of chitin. It may also participate in the defense against nematodes and other pathogens. Four isoforms of Chitinase-1 have been described that are produced by alternative splicing and isoform 3, which lacks amino acids 344-372, does not possess any enzymatic activity. Chitinase-1 is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved to generate the mature form. Its chitin-binding domain is located in amino acids 417-466 and has three chitooligosaccharide binding regions (aa 7-71; 97-100; and 210-213). Elevated levels of Chitinase-1 are observed in cases of Gaucher′s disease, bronchial asthma, and atherosclerosis. (Ref.: Kanneganti, M., et al. (2012). J. Epithel. Biol. Pharmacol. 5; 1-9).

Specificity

This rabbit polyclonal antibody detects Chitinase-1 in human serum. It targets an epitope within 15 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Chitinase-1.

Application

Anti-Chitinase-1 (CHIT1), Cat. No. ABF1168, is a rabbit polyclonal that detects Chitinase-1 in human cells and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Chitinase-1 (CHIT1) in human spleen and bone marrow tissue sections.
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in human serum.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Chitinase-1 in human serum.

Target description

~ 52 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service