Skip to Content
Merck

Skip To

ABE868

Anti-PGC-1 alpha

from rabbit, purified by affinity chromatography

Synonym(s):

Peroxisome proliferator-activated receptor gamma coactivator 1-alpha, PPAR-gamma coactivator 1-alpha, PPARGC-1-alpha

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
4
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), porcine (based on 100% sequence homology), bovine (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PPARGC1A(10891)

General description

Peroxisome proliferator-activated receptor gamma coactivator 1-alpha (UniProt: O70343; also known as PGC-1-alpha, PPAR-gamma coactivator 1-alpha, PPARGC-1-alpha) is encoded by the Ppargc1a (also known as Pgc1, Pgc1a, Ppargc1) gene (Gene ID: 19017) in murine species. PGC-1 alpha is a member of a family of transcription coactivators that plays a central role in the regulation of cellular energy metabolism. It is a homooligomeric transcriptional coactivator for steroid receptors and nuclear receptors that greatly increases the transcriptional activity of PPARG and thyroid hormone receptor on the uncoupling protein promoter. It is expressed in muscles, liver, kidney, brown adipose tissue, heart, and brain. It can regulate key mitochondrial genes that contribute to the program of adaptive thermogenesis and is also reported to play an essential role in metabolic reprogramming in response to dietary availability through coordination of the expression of a wide array of genes involved in glucose and fatty acid metabolism. PGC-1 alpha is known to induce the expression of PPARGC1- and ESRR-induced regulator, muscle 1 (PERM1) in the skeletal muscle in an estrogen-related receptors, -alpha (ESRRA)-dependent manner. PGC-1 alpha is significantly induced in brown adipose tissue and skeletal muscle by exposure to cold and is also shown to be up-regulated in brown adipose tissue of obese leptin-deficient (ob/ob) and leptin-unresponsive (db/db) mice. PGC-1 alpha is also required for oscillatory expression of clock genes, such as ARNTL/BMAL1 and NR1D1, through the co-activation of RORA and RORC, and metabolic genes, such as PDK4 and PEPCK.
~100 kDa observed; 90.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminus region of murine PPAR-gamma coactivator 1-alpha.

Application

Anti-PGC-1 alpha, Cat. No. ABE868, is a highly specific rabbit polyclonal antibody that targets Peroxisome proliferator-activated receptor gamma coactivator 1-alpha and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected PGC-1 alpha in A431, HUVEC, and NIH/3T3 cells.
Research Category
Epigenetics & Nuclear Function

Biochem/physiol Actions

This rabbit polyclonal antibody detects PGC-1 alpha in human and murine adipose tissue. it targets an epitope within 21 amino acids from the C-terminus region.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in human brown adipose tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected PGC-1 alpha in 10 µg of human brown adipose tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB2500781ST1202SAB1411910
conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

biological source

rabbit

biological source

goat

biological source

mouse

biological source

mouse

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

purified immunoglobulin

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

UniProt accession no.

O70343

UniProt accession no.

Q9UBK2

UniProt accession no.

-

UniProt accession no.

Q9UBK2

shipped in

ambient

shipped in

dry ice

shipped in

-

shipped in

dry ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters