About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
western blot: suitable
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... SIK1(150094)
General description
Immunogen
Application
Epigenetics & Nuclear Function
Cell Cycle, DNA Replication & Repair
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 1 μg/mL of this antibody detected SIK1 in human small intestine tissue lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level 100 |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form affinity isolated antibody |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| technique(s) western blot: suitable | technique(s) immunohistochemistry: 1:50- 1:200 | technique(s) immunofluorescence: suitable, western blot: 1 μg/mL | technique(s) western blot: suitable |
| Gene Information human ... SIK1(150094) | Gene Information human ... SIK1(150094) | Gene Information human ... SIK1(150094) | Gene Information human ... SIK1(150094) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



