Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Check Cart for Availability | € 395,00 |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
immunocytochemistry: suitable, immunofluorescence: suitable, western blot: suitable
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PCBP4(57060)
General description
Immunogen
Application
Immunofluorescence Analysis: 20 μg/mL from a representative lot detected PCBP4 in HeLa cells.
Epigenetics & Nuclear Function
RNA Metabolism & Binding Proteins
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 2 μg/mL of this antibody detected PCBP4 in HeLa cell lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source rabbit | biological source rabbit | biological source rabbit | biological source mouse |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form affinity isolated antibody | antibody form IgG fraction of antiserum | antibody form affinity isolated antibody | antibody form purified immunoglobulin |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level - |
| species reactivity human | species reactivity rabbit, human | species reactivity human | species reactivity human |
| Gene Information human ... PCBP4(57060) | Gene Information human ... PCBP4(57060) | Gene Information human ... PCBP3(54039) | Gene Information human ... PCBP3(54039) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



