Sign In to View Organizational & Contract Pricing.
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IHC, WB
Citations:
2
biological source
chicken
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
rat, mouse, human
technique(s)
immunofluorescence: suitable, immunohistochemistry: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PAXIP1(22976)
General description
PAX transactivation activation domain-interacting protein (PTIP) is a ubiquitous nuclear protein composed of 5 tandem BRCA1 carboxyl-terminal (BRCT) domains. PTIP was initially identified as an interacting, transactivation protein that binds to the C-terminal end of Pax2. Since its discovery, studies have shown PTIP to bind several Pax proteins, playing a critical role in response to DNA damage. It also has activity as a phosphoserine/ phosphothreonine-specific binding module, and can discern ATM and ATR gamma irradiation induced phosphorylated substrates. PTIP is involved in early cellular development and is likely to have a role in cell proliferation as well.
~120 kDa observed.
Other isoforms may be observed in some lysates.
Other isoforms may be observed in some lysates.
Immunogen
Histidine-tagged recombinant protein corresponding to human PTIP.
Application
Anti-PTIP Antibody is a Chicken Polyclonal Antibody for detection of PTIP also known as PAX transactivation activation domain-interacting protein, protein encoded by CAG trinucleotide repeats & has been validated in WB, IHC & IF.
Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in IH (Patel, S., et al. (2007).
Immunofluorescence Analysis: A represenative lot was used by an independent laboratory in IF (Lechner, M., et al. (2000).
Immunofluorescence Analysis: A represenative lot was used by an independent laboratory in IF (Lechner, M., et al. (2000).
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Chromatin Biology
Physical form
Affinity purified
Purified chicken polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Control
A431 cell lysate
A431 cell lysate
Evaluated by Western Blot in A431 cell lysate.
Western Blot Analysis: A 1:1,000 dilution of this antibody detected PTIP in 10 µg of A431 cell lysate.
Western Blot Analysis: A 1:1,000 dilution of this antibody detected PTIP in 10 µg of A431 cell lysate.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.