Skip to Content
Merck
All Photos(2)

Key Documents

ABE438

Sigma-Aldrich

Anti-Apobec1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

C->U-editing enzyme APOBEC-1, Apolipoprotein B mRNA-editing enzyme 1, HEPR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€432.00

€432.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€432.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€432.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APOBEC1(339)

General description

Apolipoprotein B mRNA-editing enzyme 1 (Apobec1) is also known as C->U-editing enzyme APOBEC-1, and HEPR. Apobec1 is a member of the cytidine and deoxycytidylate deaminase family that is exclusively expressed in the small intestine. Apobec1 is a catalytic component of the apolipoprotein B mRNA editing enzyme complex which is responsible for postranscriptional mRNA editing. Apobec1 is also thought to be involved in the epigenetic regulation of gene expression by participating in DNA demethylation.

Specificity

Other homologies: Mouse (79% sequence homology).

Immunogen

KLH-conjugated linear peptide corresponding to human Apobec1.

Application

Detect Apobec1 using this rabbit polyclonal antibody, Anti-Apobec1 Antibody validated for use in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:500-2,000 dilution from a representative lot detected Apobec1 in rat small intestine and human colon tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in human spleen tissue lysate.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected Apobec1 in 10 µg of human spleen tissue lysate.

Target description

~28 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human spleen tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoling Li et al.
Cancer cell, 41(8), 1427-1449 (2023-07-22)
Tumor mutational burden and heterogeneity has been suggested to fuel resistance to many targeted therapies. The cytosine deaminase APOBEC proteins have been implicated in the mutational signatures of more than 70% of human cancers. However, the mechanism underlying how cancer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service