Skip to Content
Merck
All Photos(1)

Key Documents

ABE2895M

Sigma-Aldrich

Anti-ORF0

from rabbit

Synonym(s):

ORF0 protein, Open Reading Frame 0

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€154.00
100 μL
€376.00

€154.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€154.00
100 μL
€376.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€154.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

primate

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), human (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

target post-translational modification

unmodified

General description

Open Reading Frame 0 (ORF0) is a primate-specific open reading frame in LINE-1 retrotransposons. Over 3,000 potential ORF0 loci are reported to be present in human and chimpanzee genomes. ORF0 is a predominantly nuclear protein that enhances LINE-1 mobility and contributes to retrotransposon-mediated diversity. It is a highly positively charged protein that may act by binding to nucleic acids. It displays two splice donor (SD) sites at nucleotides 106 and 191 (amino acids 35 and 64) that can act in concert with splice acceptors (SAs) in downstream genomic sequences to generate fusion proteins. ORF0 transcripts are reported to exist in induced pluripotent stem cells (iPSc) derived from primates. (Ref.: Denli, AM et al (2015). Cell 163(3): 583 593).

Specificity

This rabbit polyclonal antibody recognizes primate-specific open reading frame 0 (ORF0).

Immunogen

Epitope: unknown
KLH-conjugated linear peptides corresponding to ORF0 amino acid residues 20-34, 33-49, and 50-65.

Application

Anti-ORF0 Antibody, Cat. No. ABE2895, is a highly specific rabbit polyclonal antibody that targets ORF0 and has been tested in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected ORF0 in HEK293T cells transfected with ORF0 (Denli, A.M., et. al. (2015). Cell. 163(3):583-93).

Immunoprecipitation Analysis: A representative lot detected ORF0 in HEK293T cells transfected with ORF0 (Denli, A.M., et. al. (2015). Cell. 163(3):583-93).

Immunocytochemistry Analysis: A representative lot detected ORF0 in HEK293T cells transfected with ORF0 (Denli, A.M., et. al. (2015). Cell. 163(3):583-93).

Quality

Evaluated by Western Blotting in extracts from HEK293T cells transfected with ORF0 expression plasmid.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected ORF0 in lysate from HEK293T cells transfected with ORF0 expression plasmid.

Target description

~16 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service