Skip to Content
Merck
All Photos(3)

Key Documents

ABE2864M

Sigma-Aldrich

Anti-Meis1

from rabbit

Synonym(s):

Homebox protein Meis1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€154.00
100 μG
€376.00

€154.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€154.00
100 μG
€376.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€154.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

ChIP: suitable (ChIP-chip)
immunofluorescence: suitable
western blot: suitable

isotype

IgG

target post-translational modification

unmodified

Gene Information

human ... MEIS1(4211)
mouse ... Meis1(17268)

General description

Homeobox protein Meis1 (UniProt: Q60954-1; P97367-2 and P97367-3; also known as Myeloid ecotropic viral integration site 1 (Meis1) and Meis1-related protein 1) is encoded by the Meis1 gene (Gene ID: 17268). Meis proteins act as transcription factors that are orthologous to the Drosophila homothorax (Hth) protein. Meis protein contain a TALE (three-amino-acid loop extension) sub-class of the homeodomain that binds to DNA. Meis protein directly bind to Pbx proteins and the Meis/Pbx protein complex binds to DNA through respective Meis- and Pbx-consensus binding sites to regulate transcription. The Meis/Pbx complex plays important roles during development of multiple organs. In humans and mice, three homologues Meis1, Meis2 and Meis3 have been identified. Meis1 is expressed at high levels in the lung and at lower levels in the heart and brain. On the other hand, Meis 2 displays spatially restricted expression patterns in the developing nervous system, limbs, face, and in various viscera. In adult, it is mainly expressed in the brain and female genital tract, with a different distribution of the alternative splice forms in these organs. Pancreatic beta cells express both Meis1 and 2. Both Meis1 and 2 are expressed at high levels in all stages of embryonic development of mice (day 7 to 17). Deficiency of Meis1 or Meis 2 can lead to embryonic lethality. (Ref.: Machon, O et al. (2015). BMC Dev. Biol. 515:40).

Specificity

This rabbit polyclonal antibody detects Meis1 in human and murine cells. It targets an epitope within the C-terminal region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding 14 amino acids from the C-terminal region of Meis1.

Application

Anti-Meis1, Cat. No. ABE2864, is a highly specific rabbit polyclonal antibody that targets Homeobox protein Meis1 has been tested in Chromatin Immunoprecipitation (ChIP), Immunofluorescence, and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 1 µg/mL of this antibody detected Meis1 in 10 µg of K562 cell lysate.

Immunofluorescence Analysis: A representative lot detected Meis1 in adjacent stage (St) 41 sagittal hindbrain and eye sections (Mercader, N., et. al. (2005). Development. 132(18):4131-42).

Western Blotting Analysis: A 1:800 dilution from a representative lot detected Meis1 in HEK293T cells transfectd with Meis1a (Courtesy of Dr.Valeria Azcoitia and Dr. Miguel Torres).

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Meis1 in Chromatin Immunoprecipitation applications (Penkov, D., et. al. (2013). Cell Rep. 3(4):1321-33).

Western Blotting Analysis: A representative lot detected Meis1 in proximal, distal, and RA-treated distal blastemas (Mercader, N., et. al. (2005). Development. 132(18):4131-42).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Meis1 in 10 µg of HeLa cell lysate.

Target description

~52 kDa observed; 43.0 kDa (Meis1) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide and 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service