Skip to Content
Merck
All Photos(2)

Key Documents

ABE2598M

Sigma-Aldrich

Anti-CLOCK

from guinea pig

Synonym(s):

mCLOCK, CLOCK, Circadian locomoter output cycles protein kaput

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€154.00
200 μL
€376.00

€154.00


Estimated to ship on29 May 2025



Select a Size

Change View
25 μL
€154.00
200 μL
€376.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€154.00


Estimated to ship on29 May 2025


biological source

guinea pig

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

technique(s)

ChIP: suitable (ChIP-seq)
immunocytochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Clock(12753)

General description

Circadian locomoter output cycles protein kaput (UniProt: O08785; also known as mCLOCK; EC: 2.3.1.48) is encoded by the Clock gene (Gene ID: 12753) in murine species. CLOCK acts as a transcriptional activator that forms a core component of the circadian clock. CLOCK contains 2 PAS domains (aa 107-177 Pas1 domain; aa 262-332 Pas2 domain) and has a nuclear localization signaling sequence (aa 32-47). The major components of circadian clock include CRY proteins, CLOCK or NPAS2, ARNTL/BMAL1 or ARNTL2/BMAL2, CSNK1D and/or CSNK1E, TIMELESS and the PER proteins. Transcription and translation of core clock components play a critical role in rhythm generation, whereas delays imposed by post-translational modifications are important for determining the period (tau) of the rhythms. CLOCK forms a heterodimer with ARNTL/BMAL1 and this heterodimer is required for E-box-dependent transactivation and for CLOCK nuclear translocation and degradation. It is also essential for phosphorylation of both CLOCK and ARNTL/BMAL1. CLOCK has an intrinsic acetyltransferase activity, which enables circadian chromatin remodeling by acetylating histones and non-histone proteins, including ARNTL/BMAL1. CLOCK or NPAS2 and ARNTL/BMAL1 or ARNTL2/BMAL2 are reported to form the positive limb of the feedback loop, and as a heterodimer activate the transcription of core clock genes and clock-controlled genes. PER1/2/3 and CRY1/2 act as transcriptional repressors and form the negative limb of the feedback loop and inhibit transcription activation. CLOCK is expressed equally in brain, eye, testes, ovaries, liver, heart, lung, and kidney. In the brain, expression is abundant in the suprachiasmatic nuclei, pyriform cortex, and in the hippocampus.

Specificity

This guinea pig polyclonal antibody detects Circadian locomoter output cycles protein kaput (CLOCK) in human and murine cells. It targets an epitope within 39 amino acids from the internal region.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 39 amino acids from the internal region of murine Circadian locomoter output cycles protein kaput (CLOCK).

Application

Anti-CLOCK, Cat. No. ABE2598, is a highly specific guinea pig polyclonal antibody that targets Circadian locomoter output cycles protein kaput and has been tested in Chromatin Immunoprecipitation (ChIP). ChIP-seq, and Immunocytochemistry.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Circadian CLOCK in A431, HUVEC, and NIH/3T3 cells.

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected CLOCK in beta-TC6 cells (Perelis, M., et. al. (2015). Science. 350(6261):aac4250).

ChIP-seq Analysis: A representative lot detected CLOCK in beta-TC6 cells (Perelis, M., et. al. (2015). Science. 350(6261):aac4250).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected CLOCK in HeLa cells.

Target description

96.39 kda calculated.

Physical form

Affinity Purified
Format: Purified
Purified guinea pig polyclonal antibody in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service