Skip to Content
Merck
All Photos(1)

Key Documents

ABE1878

Sigma-Aldrich

Anti-UPF3A

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€130.00
100 μG
€326.00

€130.00


Estimated to ship on30 March 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€130.00
100 μG
€326.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC (p)
WB
clone:
polyclonal
species reactivity:
human
citations:
technique(s):
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

€130.00


Estimated to ship on30 March 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 54.70 kDa
observed mol wt ~54 kDa

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... UPF3A(65110)

General description

Regulator of nonsense transcripts 3A (UniProt: Q9H1J1; also known as Nonsense mRNA reducing factor 3A, Up-frameshift suppressor 3 homolog A, hUpf3, UPF3A) is encoded by the UPF3A (also known as RENT3A, UPF3) gene (Gene ID: 65110) in human. UPF3A acts as an inhibitor of nonsense-mediated decay (NMD) of mRNAs containing premature stop codons by associating with the nuclear exon junction complex (EJC) and serving as link between the EJC core and NMD machinery. It is reported to shuttle between the nucleus and the cytoplasm. It recruits UPF2 at the cytoplasmic side of the nuclear envelope and the subsequent formation of an UPF1-UPF2-UPF3 surveillance complex is believed to activate NMD. However, UPF3A is shown to be only marginally active in NMD as compared to UPF3B. It binds spliced mRNA upstream of exon-exon junctions. Mice lacking UPF3A exhibit hyper NMD and show defects in embryogenesis and gametogenesis. In human three isoforms of UPF3A have been described that are produced by alternative splicing. (Ref.: Shum, EY., et al. (2016). Cell. 165(2); 382-395).

Specificity

This rabbit polyclonal antibody detects human Regulator of nonsense transcripts 3A. It targets an epitope within 16 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal half of human Regulator of nonsense transcripts 3A (UPF3A).

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected UPF3A in HEK293 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:500 dilution from a representative lot detected UPF3A in human fetal brain tissue lysate.

Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected UPF3A in human testis and human colon tissue sections.

Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected UPF3A in lysate from HeLa cells over expressing UPF3A. (Courtesy of Matthew Kim & Dr. Wilkinson Miles at UCSD).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-UPF3A, Cat. No. ABE1878, is a rabbit polyclonal antibody that detects Regulator of nonsense transcripts 3A (UPF3A) and is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Physical form

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service