Skip to Content
Merck
All Photos(3)

Key Documents

ABE1800

Sigma-Aldrich

Anti-La-Related Protein 1

from rabbit, purified by affinity chromatography

Synonym(s):

La ribonucleoprotein domain family member 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€116.00

€116.00


Estimated to ship on07 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€116.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€116.00


Estimated to ship on07 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LARP1(23367)

Related Categories

General description

La-related protein 1 (UniProt: Q6PKG0; also known as La ribonucleoprotein domain family member 1) is encoded by the LARP1 (also known as KIAA0731, LARP) gene (Gene ID: 23367) in human. La-related protein 1 is a cytoplasmic RNA-binding protein that promotes translation of specific classes of mRNAs downstream of the mTORC1 complex. It contains 1 HTH La-type RNA-binding domain and it associates with the mRNA 5′cap in an mTOR-dependent manner and associates with mRNAs containing a 5′ terminal oligopyrimidine (5′TOP) motif, which is present in mRNAs encoding for ribosomal proteins and several components of the translation machinery. La-related protein 1 is also shown to associates with actively translating ribosomes via interaction with polyadenylate-binding protein 1 (PABPC1/PABP) and stimulates translation of mRNAs containing a 5′TOP, thereby regulating cell growth and proliferation. It is shown to be up-regulated in a number of hepatocellular carcinoma cell lines and liver cancer lesions, as well as in patients with hepatocellular carcinoma with a lower survival rate. (Ref.: Fonseca, BD et al. (2015). J. Biol. Chem. 290(26):15996-6020).

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal half of human La-related protein 1.

Application

Anti-La-Related Protein 1 Antibody, Cat. No. ABE1800, is a highly specific rabbit polyclonal antibody that targets La-related protein 1 and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected La-Related Protein 1 in HEK293 cells.

Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected La-Related Protein 1 in human cerebellum, human tonsil, and human pancreas tissues.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected La-Related Protein 1 in 10 µg of HEK293 cell lysate.

Target description

~123 kDa observed; 123.51 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service