Skip to Content
Merck

Skip To

ABE1387

Anti-Nuclear factor 1/C Antibody

serum, from rabbit

Synonym(s):

Nuclear factor 1 C-type, NF1-C, CCAAT-box-binding transcription factor, CCAAT-box-binding transcription factor, CTF, Nuclear factor I/C, NF-I/C, NFI-C, TGGCA-binding protein

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, EMSA, WB
Citations:
5


biological source

rabbit

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

ChIP: suitable, electrophoretic mobility shift assay: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NFIC(4782)

General description

Through evolution, eukaryotic cells have incorporated numerous viral promoter elements and incorporated them as part of their own gene regulatory elements. Nuclear factor 1/C was originally recognized as a transcription factor that bound the origin of replication for adenovirus type 2. Nuclear factor 1/C belongs to the CTF/NF-I family of eukaryotic DNA replication and transcription factors and is important in regulating the expression of many cellular developmental systems such as that of tooth development as well as multiple TGF-beta1 signaling mediated pathways systems.
~45-55 kDa observed

Immunogen

Epitope: C-terminus
Recombinant protein corresponding to full length Human NFIC.

Application

Chromatin Immunoprecipitation Assay (ChIP): A representative lot of this antibody has been reported to work in ChIP. See Hebbar, P.B., and Archer, T.K. (2007), Vicent G.P. et al. (2010), and Brun, M., et al. (2013).

Electrophoretic Mobility Shift Assay (EMSA): A representative lot of this antibody has been reported to work in EMSA. See Johansson E.M., et al. (2003) and Kannius-Janson, M., et al. (2002).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
This Anti-Nuclear factor 1/C Antibody is validated for use in Western Blotting and Chromatin Immunoprecipitation (ChIP) and Electrophoretic Mobility Shift Assay for the detection of Nuclear factor 1/C.

Biochem/physiol Actions

Recognizes multiple isoforms of Nuclear factor 1/C

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in U251 cell lysate.

Western Blotting Analysis: A 1:1000 dilution of this antibody detected NFIC isoforms at ~45-55 kDa in a U251 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters