Skip to Content
Merck
All Photos(2)

Documents

ABE103

Sigma-Aldrich

Anti-SIRT7 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

NAD-dependent deacetylase sirtuin-7, SIR2-like protein 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SIRT7(51547)

General description

The silent information regulator (SIR) genes (sirtuins) are a highly conserved family of proteins, which contain one or more sirtuins present in various species from bacteria to mammals. There are seven sirtuin genes that encode seven sirtuin enzymes, SIRT1 through SIRT7, all of which are dependent on oxidized nicotinamide adenine dinucleotide (NAD+). These proteins serve as mono-ADP-ribosyltransferases and deacetylate histones as a means of silencing specific chromatin regions. Further research is still required to elicidate the link between its effects on cellular metabolism, stress resistance, and cellular proliferation and its proposed role in mediating general lifespan. It is reported that Sirt7 expression demonstrates an anti-proliferative role with tumorigenic potential, in several murine cell lines.

Specificity

This antibody recognizes the N-terminus of the expressed protein of SIRT7.

Immunogen

Epitope: N-terminus.
KLH-conjugated linear peptide corresponding to the N-terminus of the expressed protein of human SIRT7.

Application

Immunohistochemistry Analysis: A 1:00 dilution from a representative lot detected the expressed protein of SIRT7 in human liver tissue lysate.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
Use Anti-SIRT7 Antibody (Rabbit Polyclonal Antibody) validated in WB, IHC to detect SIRT7 also known as NAD-dependent deacetylase sirtuin-7, SIR2-like protein 7.

Quality

Evaluated by Western Blot in human liver tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected the expressed protein of SIRT7 in 10 µg of human liver tissue lysate.

Target description

~45 kDa observed. An additional isoform at ~20 kDa is known to exist. An uncharacterized band at ~30 kDa has been observed in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human liver tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongyan Yu et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(13), 3434-3445 (2014-04-29)
Sirtuins play an important role in cancer development. Sirt7, as a member of this family, is frequently overexpressed in certain carcinomas, but the oncogenic mechanism is seldom reported. In this study, Sirt7 was characterized for its role in colorectal cancer
Guo Li et al.
The EMBO journal, 39(18), e104365-e104365 (2020-07-23)
Hair follicle stem cells (HFSCs) are maintained in a quiescent state until activated to grow, but the mechanisms that reactivate the quiescent HFSC reservoir are unclear. Here, we find that loss of Sirt7 in mice impedes hair follicle life-cycle transition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service