Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, mouse
species reactivity (predicted by homology)
primate (based on 100% sequence homology)
concentration
0.5 mg/mL
technique(s)
immunocytochemistry: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... LRIF1(55791)
General description
Immunogen
Application
Epigenetics & Nuclear Function
Chromatin Biology
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: A 1:2,000 dilution of this antibody detected Ligand-dependent nuclear receptor-interacting factor 1 in 10 µg of SH-SY5Y cell lysate.
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source rabbit | biological source rabbit | biological source rabbit | biological source mouse |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form purified immunoglobulin |
| clone polyclonal | clone polyclonal | clone polyclonal | clone H1alpha67, monoclonal |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



