Skip to Content
Merck
All Photos(3)

Documents

ABE1000

Sigma-Aldrich

Anti-FOXM1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Forkhead box protein M1, Forkhead-related protein FKHL16, Hepatocyte nuclear factor 3 forkhead homolog 11, HFH-11, HNF-3/fork-head homolog 11, M-phase phosphoprotein 2, MPM-2 reactive phosphoprotein 2, Transcription factor Trident, Winged-helix factor fr

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

canine (based on 100% sequence homology), monkey (based on 100% sequence homology), primate (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Forkhead box M1 (FOXM1), also named HNF-3, HFH-11 or Trident, belongs to a superfamily of Fox transcription factors. FOXM1 is strongly associated with cell proliferation and plays an important role in cell cycle progression by regulating both G1/S and G2/M phases. FOXM1 is also involved in DNA damage repair, angiogenesis, apoptosis, and tissue regeneration. Recently, crucial roles of this transcription factor were found in the development and progression of many cancers including colorectal, lung, prostate, liver and breast cancer. FOXM1 levels are significantly higher and associated with tumor grade in most human tumors, rendering it a potential target of cancer diagnosis and therapies.

Specificity

This antibody is expected to detect three known isoforms of FOXM1 based upon immunogen design.

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to a region near the C-terminus of human FOXM1.

Application

Anti-FOXM1 Antibody is an antibody against FOXM1 for use in western blotting, IHC & IP.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected FOXM1 in human testis tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in human thymus tissue lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected FOXM1 in 10 µg of human thymus tissue lysate.

Target description

~88 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

KLF4 is a therapeutically tractable brake on fibroblast activation that promotes resolution of pulmonary fibrosis.
Penke, et al.
JCI insight, 7 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service