Skip to Content
Merck

Skip To

ABD71

Anti-FOXC1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Forkhead box protein C1, Forkhead-related protein FKHL7, Forkhead-related transcription factor 3, FREAC-3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 452,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
1

€ 452,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXC1(2296)

General description

Forkhead box protein C1 (FOXC1) is a member of the family of forkhead transcription factors. These proteins are characterized by the forkhead domain--a 110 amino acid domain that binds to DNA molecules. FOXC1 has been implicated in the development of the eye; disruption of the FOXC1 gene has been linked to a number of glaucoma-related diseases such as Axenfeld-Rieger syndrome.
~75 kDa observed. Uniprot describes a molecular weight at ~57 kDa This protein may be observed at ~65 kDa in some cell lysates. (Tamimi, Y., et al. (2006). Hum Mol Genet. 15(21): 3229-3240.).

Immunogen

KLH-conjugated linear peptide corresponding to human FOXC1.

Application

Anti-FOXC1 Antibody is a Rabbit Polyclonal Antibody for detection of FOXC1 also known as Forkhead box protein C1, Forkhead-related protein FKHL7, FREAC-3 & has been validated in WB, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western Blot Analysis: A representative lot of this antibody detected FOXC1 in HeLa, HEK293, THP1, and MDA-MB-468 cell lysates, and in human bone marrow tissue lysate.

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected FOXC1 in HeLa cells. This antibody positively stains the nucleus, with some cytoplasm staining. This staining pattern has also been observed by an independent laboratory (Berry, F. B., et al. (2002). J Biol Chem. 277(12):10292-10297.).

Biochem/physiol Actions

Other homologies: Rat (71% sequence homology).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
THP cell lysate
Evaluated by Western Blot in THP1 cell lysate.

Western Blot Analysis: 0.025 µg/mL of this antibody detected FOXC1 in 10 µg of THP1 cell lysate. No Tween® reagent was used in the wash buffers and in the primary and secondary antibody dilution buffers.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB2277MABD79SAB4500922
description

from rabbit, purified by affinity chromatography

description

from rabbit, purified by affinity chromatography

description

clone 3G11.2, from mouse

description

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

species reactivity

human

species reactivity

rhesus macaque, rat, mouse, human

species reactivity

mouse, rat, human

species reactivity

mouse, human

Gene Information

human ... FOXC1(2296)

Gene Information

human ... FOXP1(27086)

Gene Information

human ... FOXG1(2290)

Gene Information

human ... FOXI1(2299)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters