Skip to Content
Merck
All Photos(1)

Key Documents

ABD132

Sigma-Aldrich

Anti-PHOX2B Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Paired-like homeobox 2B, Paired mesoderm homeobox protein 2B, PMX2B, Neuroblastoma Phox, NBLST2, NBPhox, PHOX2B homeodomain protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€263.00

€263.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€263.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€263.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PHOX2B(8929)

General description

Paired mesoderm homeobox protein 2B (PHOX2B), also known as Neuroblastoma Phox (NBPhox), PHOX2B homeodomain protein, and Paired-like homeobox 2B is a paired-homeodomain transcription factor that maintains the noradrenergic phenotype during embryogenesis. PHOX2 proteins help regulate endogenous hydroxylases in neural crest cells and aid in sympathetic neuron generation. PHOX2A controls only the differentiation of the main noradrenergic center of the human brain, while PHOX2B is required for the differentiation of all central and nonperipheral noradrenergic centers. PHOX2B is expressed in neuroblastoma, brain and adrenal gland. PHOX2B defects can cause congenital central hypoventilation syndrome (CCHS) and susceptibility to neuroblastoma type 2 (NBLST2). PHOX2 regulation may have therapeutic potential in aging or degenerative noradrenergic neuron disorders.

Immunogen

KLH-conjugated linear peptide corresponding to human PHOX2B.

Application

Detect Phox2a using this rabbit polyclonal antibody, Anti-PHOX2B Antibody validated for use in western blotting.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected PHOX2B in 15 µg of HEK293 cell lysate.

Target description

~22 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service