Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, mouse
species reactivity (predicted by homology)
rat (based on 100% sequence homology)
packaging
antibody small pack of 25 μg
technique(s)
immunohistochemistry: suitable (paraffin), western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... MUSTN1(389125)
General description
Immunogen
Application
Cell Structure
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected MUSTN1 in human skeletal muscle, mouse embryo, and mouse embryonic lung tissues.
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected MUSTN1 in human skeletal muscle, mouse embryo, and mouse embryonic lung tissues.
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 2 µg/mL of this antibody detected MUSTN1 in C2C12 cell lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description from rabbit, purified by affinity chromatography | description IgG fraction of antiserum | description clone AD6-1, from mouse | description clone 15D5.1, from mouse |
| biological source rabbit | biological source rabbit | biological source mouse | biological source mouse |
| Quality Level 100 | Quality Level - | Quality Level 100 | Quality Level 100 |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form affinity isolated antibody | antibody form IgG fraction of antiserum | antibody form purified antibody | antibody form purified antibody |
| clone polyclonal | clone polyclonal | clone AD6-1, monoclonal | clone 15D5.1, monoclonal |
| technique(s) western blot: suitable, immunohistochemistry: suitable (paraffin) | technique(s) western blot: 1:500-1:2,000 | technique(s) immunocytochemistry: suitable, western blot: suitable | technique(s) immunohistochemistry: suitable (paraffin), western blot: suitable |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
wgk
WGK 1
Storage Class
12 - Non Combustible Liquids
flash_point_f
does not flash
flash_point_c
does not flash
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



