Skip to Content
Merck
All Photos(1)

Key Documents

ABC961

Sigma-Aldrich

Anti-OFD1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Oral-facial-digital syndrome 1 protein homolog, OFD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

The cilium is a cell surface projection found in many vertebrate cells and it is required to transduce signals important for development and tissue homeostasis. OFD1 is important in cilium development, and thus it is important in cellular development and signaling. OFD1 is a component of the centrioles and appears to control the length of the centrioles in mother-daughter cells. OFD1 also plays an important role in the WNT signaling pathway and in the formation of the left-right axis in embryos. OFD1 is expressed in the cytoplasm during development in most tissues but can be found in the nucleus as well, and it is known to interact with proteins important in chromatin remodeling as well. At least one X-linked genetic mutation is associated with defects in the OFD1 gene. It is a form of oroffaciodigital syndrome characterized by abnormalities in the oral cavity, face, digits and other disfigurements that is lethal in males.

Immunogen

Epitope: C-terminus
Linear peptide corresponding to the C-terminus of mouse OFD1.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-OFD1 Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of OFD1.
Western Blotting Analysis: A representative lot detected OFD1 in untreated and autophagy-induced/serum-starved mouse MEFs (Tang, Z., et al. (2013). Nature. 502:254-257).
Western Blotting Analysis: A representative lot detected OFD1 in wild-type female mouse embryo lysates (Ferrante, M.I., et al. (2006). Nat Genet. 38(1):112-117).
Western Blotting Analysis: A representative lot detected OFD1 in male E10.5 knock-out mouse embryo lysates (Ferrante, M.I., et al. (2006). Nat Genet. 38(1):112-117).
Immunocytochemistry Analysis: A representative lot detected OFD1 in PCM1 knockdown U2OS cells & untreated and serum-starved/autophagy-induced U2OS cells where OFD1 partially colocalizes with LC3 (Tang, Z., et al. (2013). Nature. 502:254-257).

Quality

Evaluated by Western Blotting in U2OS cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected OFD1 in 10 µg of U2OS cell lysate.

Target description

~120 kDa observed. Uncharacterized bands may be observed in some lysates at 45 kDa, 33 kDa & 28 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noémie Gaudin et al.
eLife, 11 (2022-03-24)
Centrioles are formed by microtubule triplets in a ninefold symmetric arrangement. In flagellated protists and animal multiciliated cells, accessory structures tethered to specific triplets render the centrioles rotationally asymmetric, a property that is key to cytoskeletal and cellular organization in
Hideki Izumi et al.
Scientific reports, 9(1), 2236-2236 (2019-02-21)
CD133 is a transmembranous protein that mainly localises to the plasma membrane in haematopoietic and neural stem cells as well as cancer stem cells. Although CD133 also localises to the cytoplasm, the mechanism of action and function of cytoplasmic CD133
Emanuela Senatore et al.
The EMBO journal, 40(10), e106503-e106503 (2021-05-03)
The primary cilium is a microtubule-based sensory organelle that dynamically links signalling pathways to cell differentiation, growth, and development. Genetic defects of primary cilia are responsible for genetic disorders known as ciliopathies. Orofacial digital type I syndrome (OFDI) is an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service