Skip to Content
Merck
All Photos(2)

Documents

ABC937

Sigma-Aldrich

Anti-FOXL1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Forkhead box protein L1, Forkhead-related protein FKHL11, Forkhead-related transcription factor 7, FREAC-7, FOXL1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

sheep (based on 100% sequence homology), feline (based on 100% sequence homology), porcine (based on 100% sequence homology), bovine (based on 100% sequence homology), horse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXL1(2300)

General description

Forkhead box protein L1 (UniProt Q12952; also known as Forkhead-like 11, Forkhead-related protein FKHL11, Forkhead-related transcription factor 7, FREAC-7) is encoded by the FOXL1 (also known as FKHL11, FREAC7) gene (Gene ID 2300) in human. FOXL1 protein belongs to the forehead box (Fox) family of transcription factors characterized by a common DNA-binding domain (the forkhead box). FOXL1 is involved in regulating the proliferation of gastrointestinal epithelial cells. Loss of Foxl1 leads to distorted architecture of the stomach and small intestine in mice due to a markedly increased epithelial proliferation. Foxl1 knockout is correlated with an enhanced Wnt/β-catenin pathway activation as evidenced by increased β-catenin nuclear localization. FOXL1 expression is down-regulated in the majority of human clear cell renal cell carcinoma (ccRCC), at both mRNA and protein levels. FOXL1 expression inversely correlates with the aggressive phenotype of ccRCC and the survival outcome of patients.

Immunogen

KLH-conjugated linear peptide corresponding to human FOXL1.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected FOXL1 in human breast cancer tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-FOXL1 Antibody is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of FOXL1.

Quality

Evaluated by Western Blotting in human colon tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected FOXL1 in 10 µg of human colon tissue lysate.

Target description

~36 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroki Kobayashi et al.
Gastroenterology, 160(4), 1224-1239 (2020-11-17)
Cancer-associated fibroblasts (CAFs), key constituents of the tumor microenvironment, either promote or restrain tumor growth. Attempts to therapeutically target CAFs have been hampered by our incomplete understanding of these functionally heterogeneous cells. Key growth factors in the intestinal epithelial niche

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service